Search
Search

Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
13 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|
||||||||||||||||||||||||||||||||||||||
Literature Links: |
CASP9 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
| 1000Genome | Applied Biosystems® | HapMap | |||
|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
| CASP9 - caspase 9 | ||||||
|---|---|---|---|---|---|---|
| Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
| NM_001229.4 | 484 | Silent Mutation | ATC,ATG | I,M 185 | NP_001220.2 | |
| NM_001278054.1 | 484 | Intron | NP_001264983.1 | |||
| NM_032996.3 | 484 | Silent Mutation | ATC,ATG | I,M 102 | NP_127463.2 | |
| XM_005246014.2 | 484 | Silent Mutation | ATC,ATG | I,M 102 | XP_005246071.1 | |
| XM_011542273.2 | 484 | Missense Mutation | ATC,ATG | I,M 185 | XP_011540575.1 | |