Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGGCATGAACCATCACGCCTGACCA[A/T]GTTCCAGATTTGTAACTTGGTCATT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605563 MIM: 613802 | ||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CABP1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
CABP1 - calcium binding protein 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MLEC - malectin | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001303627.1 | 2007 | Intron | NP_001290556.1 | |||
NM_001303628.1 | 2007 | Intron | NP_001290557.1 | |||
NM_014730.3 | 2007 | Intron | NP_055545.1 | |||
XM_011539031.1 | 2007 | UTR 5 | XP_011537333.1 | |||
XM_011539032.1 | 2007 | UTR 5 | XP_011537334.1 |
Set Membership: |
HapMap |