Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604113 MIM: 164009 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
IL18BP PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
IL18BP - interleukin 18 binding protein | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001039659.1 | 7093 | Intron | NP_001034748.1 | |||
NM_001039660.1 | 7093 | Intron | NP_001034749.1 | |||
NM_001145055.1 | 7093 | Intron | NP_001138527.1 | |||
NM_001145057.1 | 7093 | Intron | NP_001138529.1 | |||
NM_005699.3 | 7093 | Intron | NP_005690.2 | |||
NM_173042.2 | 7093 | Intron | NP_766630.2 | |||
NM_173044.2 | 7093 | Intron | NP_766632.2 | |||
XM_017017059.1 | 7093 | Intron | XP_016872548.1 | |||
XM_017017060.1 | 7093 | Intron | XP_016872549.1 | |||
XM_017017061.1 | 7093 | Intron | XP_016872550.1 | |||
XM_017017062.1 | 7093 | Intron | XP_016872551.1 | |||
XM_017017063.1 | 7093 | Intron | XP_016872552.1 |
NUMA1 - nuclear mitotic apparatus protein 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001286561.1 | 7093 | UTR 3 | NP_001273490.1 | |||
NM_006185.3 | 7093 | UTR 3 | NP_006176.2 | |||
XM_006718564.1 | 7093 | UTR 3 | XP_006718627.1 | |||
XM_011545054.1 | 7093 | UTR 3 | XP_011543356.1 | |||
XM_011545055.1 | 7093 | UTR 3 | XP_011543357.1 | |||
XM_011545056.2 | 7093 | UTR 3 | XP_011543358.1 | |||
XM_011545057.2 | 7093 | UTR 3 | XP_011543359.1 | |||
XM_011545058.1 | 7093 | UTR 3 | XP_011543360.1 | |||
XM_011545059.2 | 7093 | UTR 3 | XP_011543361.1 | |||
XM_011545060.2 | 7093 | UTR 3 | XP_011543362.1 | |||
XM_011545061.2 | 7093 | UTR 3 | XP_011543363.1 | |||
XM_011545062.2 | 7093 | UTR 3 | XP_011543364.1 | |||
XM_011545063.2 | 7093 | UTR 3 | XP_011543365.1 | |||
XM_011545064.1 | 7093 | UTR 3 | XP_011543366.1 | |||
XM_011545065.1 | 7093 | UTR 3 | XP_011543367.1 | |||
XM_011545066.1 | 7093 | UTR 3 | XP_011543368.1 | |||
XM_017017830.1 | 7093 | Intron | XP_016873319.1 | |||
XM_017017831.1 | 7093 | Intron | XP_016873320.1 |
RNF121 - ring finger protein 121 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap |