Search
Search

ELF2TATTATTAAGTGCAAACTACCCTCA[A/C]CCCTTTAAGGATTTTCTTATCATTT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
ELF2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
| 1000Genome | Applied Biosystems® | HapMap |
|---|---|---|
| Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
| EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
| SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
| AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
| EUR - Not Available | ||
| AMR - Not Available |
| ELF2 - E74 like ETS transcription factor 2 | ||||||
|---|---|---|---|---|---|---|
| Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
| NM_001276457.1 | Intron | NP_001263386.1 | ||||
| NM_001276458.1 | Intron | NP_001263387.1 | ||||
| NM_001276459.1 | Intron | NP_001263388.1 | ||||
| NM_006874.3 | Intron | NP_006865.1 | ||||
| NM_201999.2 | Intron | NP_973728.1 | ||||
| XM_005262803.2 | Intron | XP_005262860.1 | ||||
| XM_005262804.2 | Intron | XP_005262861.1 | ||||
| XM_005262805.2 | Intron | XP_005262862.1 | ||||
| XM_006714128.2 | Intron | XP_006714191.1 | ||||
| XM_006714129.3 | Intron | XP_006714192.1 | ||||
| XM_011531710.2 | Intron | XP_011530012.1 | ||||
| XM_011531711.2 | Intron | XP_011530013.1 | ||||
| XM_011531712.2 | Intron | XP_011530014.1 | ||||
| XM_011531714.2 | Intron | XP_011530016.1 | ||||
| XM_017007856.1 | Intron | XP_016863345.1 | ||||
| XM_017007857.1 | Intron | XP_016863346.1 | ||||
| XM_017007858.1 | Intron | XP_016863347.1 | ||||
| XM_017007859.1 | Intron | XP_016863348.1 | ||||
| XM_017007860.1 | Intron | XP_016863349.1 | ||||
| XM_017007861.1 | Intron | XP_016863350.1 | ||||