Search
Search

ARHGAP30
USF1ATGACTGATCCTTTCAGACCAATGT[C/T]CCTAGTTATTGGCTTGCTACCTTAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
33 submissions
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ARHGAP30 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
| 1000Genome | Applied Biosystems® | HapMap | ||||||
|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
| ARHGAP30 - Rho GTPase activating protein 30 | ||||||
|---|---|---|---|---|---|---|
| Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
| NM_001025598.1 | Intron | NP_001020769.1 | ||||
| NM_001287600.1 | Intron | NP_001274529.1 | ||||
| NM_001287602.1 | Intron | NP_001274531.1 | ||||
| NM_181720.2 | Intron | NP_859071.2 | ||||
| XM_005245070.2 | Intron | XP_005245127.1 | ||||
| XM_005245073.3 | Intron | XP_005245130.1 | ||||
| XM_011509391.2 | Intron | XP_011507693.1 | ||||
| XM_017000960.1 | Intron | XP_016856449.1 | ||||
| USF1 - upstream transcription factor 1 | ||||||
|---|---|---|---|---|---|---|
| There are no transcripts associated with this gene. | ||||||
Set Membership: |
|