Search
Search

Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ABCC6 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
| 1000Genome | Applied Biosystems® | HapMap | ||||||
|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
| ABCC6 - ATP binding cassette subfamily C member 6 | ||||||
|---|---|---|---|---|---|---|
| Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
| NM_001079528.3 | Intron | NP_001072996.1 | ||||
| NM_001171.5 | Intron | NP_001162.4 | ||||
| XM_011522479.2 | Intron | XP_011520781.1 | ||||
| XM_011522480.1 | Intron | XP_011520782.1 | ||||
| XM_011522481.2 | Intron | XP_011520783.1 | ||||
| XM_011522482.2 | Intron | XP_011520784.1 | ||||
| XM_017023212.1 | Intron | XP_016878701.1 | ||||
| XM_017023213.1 | Intron | XP_016878702.1 | ||||
| XM_017023214.1 | Intron | XP_016878703.1 | ||||
Set Membership: |
|