Search
Search

| Assay Name: | cel-miR-77 |
|---|---|
| miRBase Accession Number: | MI0000048 |
| miRBase Version: | v22.1 |
| Mature miRNA Sequence: | UUCAUCAGGCCAUAGCUGUCCA |
| Species: | Caenorhabditis elegans, Caenorhabditis brenneri, Caenorhabditis briggsae |
| Product Type: | TaqMan™ MicroRNA Assay |
Gene Family ID: | MIPF0000272, mir-77 |
Mature miRNA Sequence: | UUCAUCAGGCCAUAGCUGUCCA |
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Caenorhabditis elegans | cel-miR-77-3p | MIMAT0000049 |
| Caenorhabditis brenneri | cbn-miR-77 | MIMAT0029778 |
| Caenorhabditis briggsae | cbr-miR-77 | MIMAT0000485 |