Search
Search

| Assay Name: | hsa-miR-486-3p |
|---|---|
| Stem-loop Accession Number: | MI0002470 |
| miRBase Version: | v22.1 |
| Mature miRNA Sequence: | CGGGGCAGCUCAGUACAGGAU |
| Species: | Human, Mouse, Horse, Pteropus alecto, Rhesus monkey, Tupaia chinensis |
| Product Type: | TaqMan™ Advanced miRNA Assay |
Gene Family ID: | MIPF0000220, mir-486 |
Mature miRNA Sequence: | CGGGGCAGCUCAGUACAGGAU |
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Human | hsa-miR-486-3p | MIMAT0004762 |
| Mouse | mmu-miR-486a-3p | MIMAT0017206 |
| Horse | eca-miR-486-3p | MIMAT0013187 |
| Pteropus alecto | pal-miR-486a-3p | MIMAT0039847 |
| Rhesus monkey | mml-miR-486-3p | MIMAT0006345 |
| Tupaia chinensis | tch-miR-486-3p | MIMAT0036553 |