Search
Search

| Assay Name: | sme-miR-750-5p |
|---|---|
| miRBase Accession Number: | MI0005176 |
| miRBase Version: | v22.1 |
| Mature miRNA Sequence: | GCACUGGAAGGUUUGGUUUAGCU |
| Species: | Schmidtea mediterranea |
| Product Type: | TaqMan™ MicroRNA Assay |
Mature miRNA Sequence: | GCACUGGAAGGUUUGGUUUAGCU |
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Schmidtea mediterranea | sme-miR-750-5p | MIMAT0012124 |