Search
Search

| Assay Name: | tgu-miR-155 |
|---|---|
| miRBase Accession Number: | MI0029760 |
| miRBase Version: | v22.1 |
| Mature miRNA Sequence: | UUAAUGCUAAUCGUGAUAGGG |
| Species: | Alligator mississippiensis, Taeniopygia guttata |
| Product Type: | TaqMan™ MicroRNA Assay |
Mature miRNA Sequence: | UUAAUGCUAAUCGUGAUAGGG |
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Alligator mississippiensis | ami-miR-155-5p | MIMAT0038185 |
| Taeniopygia guttata | tgu-miR-155-5p | MIMAT0014612 |