Search
Search

| Assay Name: | cbn-miR-7614 |
|---|---|
| miRBase Accession Number: | MI0024793 |
| miRBase Version: | v22.1 |
| Mature miRNA Sequence: | UAACCCAACUGAAAAUAGCUGGA |
| Species: | Caenorhabditis brenneri |
| Product Type: | TaqMan™ MicroRNA Assay |
Mature miRNA Sequence: | UAACCCAACUGAAAAUAGCUGGA |
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Caenorhabditis brenneri | cbn-miR-7614 | MIMAT0029652 |