Search
Search

| Assay Name: | bta-miR-6123 |
|---|---|
| miRBase Accession Number: | MI0021141 |
| miRBase Version: | v22.1 |
| Mature miRNA Sequence: | UGCCAAGCCCACGUUCAAAGG |
| Species: | Bovine |
| Product Type: | TaqMan™ MicroRNA Assay |
Mature miRNA Sequence: | UGCCAAGCCCACGUUCAAAGG |
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Bovine | bta-miR-6123 | MIMAT0024596 |