Search
Search

| Assay Name: | hsa-miR-296-5p |
|---|---|
| Stem-loop Accession Number: | MI0000747 |
| miRBase Version: | v22.1 |
| Mature miRNA Sequence: | AGGGCCCCCCCUCAAUCCUGU |
| Species: | Human, Mouse, Rat, Callithrix jacchus, Guinea pig, Pongo pygmaeus, Rhesus monkey |
| Product Type: | TaqMan™ Advanced miRNA Assay |
Gene Family ID: | MIPF0000159, mir-296 |
Mature miRNA Sequence: | AGGGCCCCCCCUCAAUCCUGU |
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Human | hsa-miR-296-5p | MIMAT0000690 |
| Mouse | mmu-miR-296-5p | MIMAT0000374 |
| Rat | rno-miR-296-5p | MIMAT0000898 |
| Callithrix jacchus | cja-miR-296 | MIMAT0039374 |
| Guinea pig | cpo-miR-296-5p | MIMAT0047138 |
| Pongo pygmaeus | ppy-miR-296-5p | MIMAT0015809 |
| View More Rows | ||
| Rhesus monkey | mml-miR-296-5p | MIMAT0006251 |