Search
Search

| Assay Name: | hsa-miR-139-3p |
|---|---|
| Stem-loop Accession Number: | MI0000261 |
| miRBase Version: | v22.1 |
| Mature miRNA Sequence: | UGGAGACGCGGCCCUGUUGGAGU |
| Species: | Human, Cricetulus griseus, Dasypus novemcinctus, Eptesicus fuscus, Guinea pig, Pig, Pteropus alecto, Rabbit |
| Product Type: | TaqMan™ Advanced miRNA Assay |
Gene Family ID: | MIPF0000117, mir-139 |
Mature miRNA Sequence: | UGGAGACGCGGCCCUGUUGGAGU |
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Human | hsa-miR-139-3p | MIMAT0004552 |
| Cricetulus griseus | cgr-miR-139-3p | MIMAT0023766 |
| Dasypus novemcinctus | dno-miR-139-3p | MIMAT0047652 |
| Eptesicus fuscus | efu-miR-139 | MIMAT0035163 |
| Guinea pig | cpo-miR-139-3p | MIMAT0047003 |
| Pig | ssc-miR-139-3p | MIMAT0022921 |
| View More Rows | ||
| Pteropus alecto | pal-miR-139-3p | MIMAT0039926 |
| Rabbit | ocu-miR-139-3p | MIMAT0048240 |