Hamburger Menu Button
Thermo Fisher Scientific Logo
登入
還沒有帳戶嗎 ? 立即註冊
  • 產品
    • Antibodies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Greener Products
    • 查看所有產品類別
  • 應用
    • 生物製程
    • 細胞培養和轉染
    • 食品和飲料
    • 實驗室解決方案
    • 製藥和生物製藥
    • 即時定量PCR
    • 半導體分析
    • 臨床和診斷
    • 數位解決方案
    • 分子光譜、同位素與元素分析
    • 查看所有應用與技術
  • 服務
    • 360° CDMO 和 CRO 服務
    • CDMO 服务
    • 临床试验 CRO 服务
    • 客製化服務
    • 金融與租賃服務
    • 儀器服務
    • 實驗室資訊學
    • 授權與商業供應
    • 培訓服務
    • 查看所有服務
  • 支援
    • 線上訂購協助
    • 數位解決方案
    • 產品支持
    • 技術相關資訊
    • 培訓與教育
    • 查看所有協助與支援
  • 熱門商品
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • 我們的服務對象
    • 生物技術
    • 生物製藥
    • CDMO
    • 實驗室診斷
    • 工業與應用科學
  • 特價商品
  • 聯絡我們
  • 快速訂購
  • 訂單狀態與貨物追蹤
  • 文件和證書
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • 訂單查詢
          • 快速訂購
          • 登入
            登入
            還沒有帳戶嗎 ? 立即註冊
            • 帳戶
            • 訂單
            • Connect: Lab, Data, Apps
            • 客製化產品與項目
            • 儀器管理
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › 478002_mir

          Specificity testing was performed using human anti-targets.

          Assay Name: hsa-miR-29a-5p
          Stem-loop Accession Number: MI0000087
          miRBase Version: v22.1
          Chromosome Location: Chr.7: 130876747 - 130876810 [-] on Build GRCh38
          Mature miRNA Sequence: ACUGAUUUCUUUUGGUGUUCAG
          Species: Human, Mouse, Rat, Ornithorhynchus anatinus, Pig, Pteropus alecto
          Product Type: TaqMan™ Advanced miRNA Assay
          Assay ID 478002_mir
          Size S: 250 rxns
          Availability Inventoried
          Catalog # A25576
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Family ID:

          MIPF0000009, mir-29

          Mature miRNA Sequence:

          ACUGAUUUCUUUUGGUGUUCAG

          miRBase Details:



          Species miRBase ID miRBase Accession Number
          Human hsa-miR-29a-5p MIMAT0004503
          Mouse mmu-miR-29a-5p MIMAT0004631
          Rat rno-miR-29a-5p MIMAT0004718
          Ornithorhynchus anatinus oan-miR-29a-1-5p MIMAT0006806
          Pig ssc-miR-29a-5p MIMAT0037364
          Pteropus alecto pal-miR-29a-5p MIMAT0039801

          Stem-loop Details



          Human

          Stem-loop ID hsa-mir-29a
          Stem-loop Accession # MI0000087
          Stem-loop Sequence
          AUGACUGAUUUCUUUUGGUGUUCAGAGUCAAUAUAAUUUUCUAGCACCAUCUGAAAUCGGUUAU
          Chromosome Location Chr. 7 - 130876747 - 130876810 [-] on Build GRCh38
          Mature MicroRNA miRBase Accession #: MIMAT0004503
          miRBase ID: hsa-miR-29a-5p
          Mature miRNA Sequence: ACUGAUUUCUUUUGGUGUUCAG
          Chromosome Location: Chr. 7 - 130876747 - 130876810 [-] on Build GRCh38

          Mouse

          Stem-loop ID mmu-mir-29a
          Stem-loop Accession # MI0000576
          Stem-loop Sequence
          ACCCCUUAGAGGAUGACUGAUUUCUUUUGGUGUUCAGAGUCAAUAGAAUUUUCUAGCACCAUCUGAAAUCGGUUAUAAUGAUUGGGGA
          Chromosome Location Chr. 6 - 31062660 - 31062747 [-] on Build GRCm38
          Mature MicroRNA miRBase Accession #: MIMAT0004631
          miRBase ID: mmu-miR-29a-5p
          Mature miRNA Sequence: ACUGAUUUCUUUUGGUGUUCAG
          Chromosome Location: Chr. 6 - 31062660 - 31062747 [-] on Build GRCm38

          Rat

          Stem-loop ID rno-mir-29a
          Stem-loop Accession # MI0000863
          Stem-loop Sequence
          ACCCCUUAGAGGAUGACUGAUUUCUUUUGGUGUUCAGAGUCAAUAGAAUUUUCUAGCACCAUCUGAAAUCGGUUAUAAUGAUUGGGGA
          Chromosome Location Chr. 4 - 58343931 - 58344018 [-] on Build Rnor_6.0
          Mature MicroRNA miRBase Accession #: MIMAT0004718
          miRBase ID: rno-miR-29a-5p
          Mature miRNA Sequence: ACUGAUUUCUUUUGGUGUUCAG
          Chromosome Location: Chr. 4 - 58343931 - 58344018 [-] on Build Rnor_6.0

          Ornithorhynchus anatinus

          Stem-loop ID oan-mir-29a-1
          Stem-loop Accession # MI0006668
          Stem-loop Sequence
          UCAAAGGCUCCUGUGACCCCUCUAGAGGAUGACUGAUUUCUUUUGGUGUUCAGAGUCAAUAUCAUUUUCUAGCACCAUUUGAAAUCGGUUAUAAUGAUUGGGGAAUAGCGCCACGUGA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0006806
          miRBase ID: oan-miR-29a-1-5p
          Mature miRNA Sequence: ACUGAUUUCUUUUGGUGUUCAG
          Chromosome Location: NA

          Pig

          Stem-loop ID ssc-mir-29a
          Stem-loop Accession # MI0013090
          Stem-loop Sequence
          CCCCUUAGAGGAUGACUGAUUUCUUUUGGUGUUCAGAGUCAAUAUAAUUUUCUAGCACCAUCUGAAAUCGGUUAUAAUGAUUGGGG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0037364
          miRBase ID: ssc-miR-29a-5p
          Mature miRNA Sequence: ACUGAUUUCUUUUGGUGUUCAG
          Chromosome Location: NA

          Pteropus alecto

          Stem-loop ID pal-mir-29a
          Stem-loop Accession # MI0032336
          Stem-loop Sequence
          ACUGAUUUCUUUUGGUGUUCAGAGUCAAUACAAUUUUCUAGCACCAUCUGAAAUCGGUUA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0039801
          miRBase ID: pal-miR-29a-5p
          Mature miRNA Sequence: ACUGAUUUCUUUUGGUGUUCAG
          Chromosome Location: NA


          Back To Top

          More Information




          Related Products

          TaqMan™ Pri-miRNA Assay : Rn03464529_pri
          TaqMan™ Pri-miRNA Assay : Mm03306859_pri
          TaqMan™ Pri-miRNA Assay : Hs03302672_pri
          Ambion® Anti-miR™ miRNA Inhibitor : AM12463
          Ambion® Pre-miR™ miRNA Precursor : PM12463
          TaqMan™ MicroRNA Assay : 002447
          mirVana® miRNA inhibitor : MH12463
          mirVana® miRNA mimic : MC12463
          TaqMan™ Advanced miRNA Assay : mmu481032_mir
          TaqMan™ Advanced miRNA Assay : rno481032_mir
          FGMIR : TaqManTM Advanced miRNA Human B 96-well Plate 3
          FGMIR : TaqManTM Advanced miRNA Human B 96-well Plate 3
          FGMIR : TaqManTM Advanced miRNA Human Serum/Plasma Card
          FGMIR : TaqManTM Advanced miRNA Human B Card
          MIRNA : TaqMan® Advanced miRNA Human Serum/Plasma 96-well Plates, Standard
          FGMIR : QS TaqmanTM OpenArray Human Advanced miRNA Panel
          MIRNA : TaqMan® Advanced miRNA Human B 96-well Plates, Standard
          MIRNA : TaqMan® OpenArray® Human Advanced MicroRNA Panel
          MIRNA : TaqMan® Advanced miRNA Human A and B 96-well Plates, Standard
          FGMIR : TaqManTM Advanced miRNA Human Serum/Plasma 96-well Plate 2
          MIRNA : TaqMan® Advanced miRNA Human Serum/Plasma 96-well Plates, Fast
          MIRNA : TaqMan® Advanced miRNA Human Serum/Plasma Card
          MIRNA : TaqMan® Advanced miRNA Human B Card
          FGMIR : TaqManTM Advanced miRNA Human Serum/Plasma 96-well Plate 2
          MIRNA : TaqMan® Advanced miRNA Human A and B Cards

          Back To Top

          Related Products

          • TaqMan® Advanced miRNA cDNA Synthesis Kit
          • TaqMan® Fast Advanced Master Mix
          產品訂購 Plus Icon Minus Icon
          • 訂單狀態
          • 訂購協助
          • 快速訂購
          • 現貨供應中心
          • 電子採購
          支持 Plus Icon Minus Icon
          • 協助與支持
          • 聯絡我們
          • 技術支持中心
          • 搜尋文件和證書
          • 回報網站問題
          資源 Plus Icon Minus Icon
          • 學習中心
          • 促銷活動
          • 活動和網路研討會
          • 社群媒體
          關於 Thermo Fisher Plus Icon Minus Icon
          • 關於我們 關於我們
          • 人才招募 人才招募
          • 投資者資訊 投資者資訊
          • 新聞 新聞
          • 企業社會責任 企業社會責任
          • 商標
          • 政策和聲明
          品牌 Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-n9r8p:80/100.66.76.150:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline