Search
Search

| Assay Name: | |
|---|---|
| Stem-loop Accession Number: | MI0007693 |
| miRBase Version: | v22.1 |
| Mature miRNA Sequence: | ACUGCCCCAGGUGCUGCUGG |
| Species: | Pan troglodytes, Pongo pygmaeus, Rhesus monkey |
| Product Type: | TaqMan™ Advanced miRNA Assay |
Gene Family ID: | MIPF0000165, mir-324 |
Mature miRNA Sequence: | ACUGCCCCAGGUGCUGCUGG |
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Pan troglodytes | ptr-miR-324 | MIMAT0008098 |
| Pongo pygmaeus | ppy-miR-324-3p | MIMAT0015828 |
| Rhesus monkey | mml-miR-324-3p | MIMAT0006267 |