Search
Search

| Assay Name: | hsa-miR-3180-5p |
|---|---|
| Stem-loop Accession Number: | MI0014214 |
| miRBase Version: | v22.1 |
| Mature miRNA Sequence: | CUUCCAGACGCUCCGCCCCACGUCG |
| Species: | Human |
| Product Type: | TaqMan™ Advanced miRNA Assay |
Gene Family ID: | MIPF0000894, mir-3180 |
Mature miRNA Sequence: | CUUCCAGACGCUCCGCCCCACGUCG |
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Human | hsa-miR-3180-5p | MIMAT0015057 |