Recent searches Clear History
Kontaktieren Sie uns
Allergen Encyclopedia
Table of Contents
Komponente
f424 Ara h 3
f424 Ara h 3 Scientific Information
Typ:
Component
Name; WHO/IUIS:
Ara h 3
Route of Exposure:
Ingestion
Biological function:
11S globulin trypsin inhibitor
Allergen code:
f424
Source Material:
Peanut extract
Other Names :
Glycinin
Summary
Ara h 3 (glycinin) is a storage protein and a cupin allergen able to withstand heat treatment and enzymatic activity. It is estimated that 97% of peanut allergy patients are sensitized to at least one of the allergens Ara h 1, 2 and 3. Individuals who are sensitized to Ara h 3 are at an increased risk for more severe symptoms and anaphylactic reactions. Polysensitization to Ara 1, 2 and 3 can help to predict the severity of reaction at challenge. The use of specific IgE to Ara h 3 in combination with Ara h 1 and 2 could be helpful to diagnose individuals with peanut allergy. Cross-reactivity of Ara h 3 has been shown with Ara h 1, other legumes and legumin tree nut allergens. 
Epidemiology
Worldwide distribution
Studies have typically reported peanut allergy prevalence rates between 1–2% in Western nations. Peanut allergy appears to be less common in Asia and other global areas, although epidemiological studies in non-Western regions have been sparse (1). Peanut allergy usually begins in childhood and persists throughout the affected individual’s lifetime however, approximately 20% of young children develop tolerance (2).
The results of a study published in 1999 found that just over half (53%) of 40 peanut-allergic patients were sensitized to Ara h 4 which is now known to be an isoform of Ara h 3, Ara h 3.02 (3, 4). In the USA, it is estimated that the prevalence is lower with just over a third of patients sensitized to Ara h 3 and 1% being monosensitized to this allergen component (5). The highest frequency of positive test results for Ara h 3 in the USA study was within the three to nine-year old group, which decreased with increasing age. Despite the lower overall prevalence of Ara h 3 sensitization in the USA, Ara h 3 is considered to be a major peanut allergen (4).
Environmental Characteristics
Source and tissue
Ara h 3 is a cupin allergen belonging to the legumin family. Legumins, also known as 11S globulins, are hexameric proteins present in the seeds of many plants (6). The allergens Ara h 1, 2 and 3 provide >30% of the total protein content of peanuts (7).
Clinical Relevance
As a storage protein, individuals who are sensitized to Ara h 3 are at an increased risk for more severe symptoms and anaphylactic reactions (8). Sensitization to peanut storage proteins Ara h 1, 2 and 3 was associated with increased quantities of airway and systemic inflammation markers compared to patients who were not sensitized to these peanut allergen components in a population of asthma sufferers (9). Food hypersensitivity symptoms were also more regularly reported by those sensitized to Ara h 1, 2 and 3 than by any other participants in the study (9).
A study found IgE was positively correlated with clinical severity and Ara h 3 in adult patients (r = 0.78, P < 0.001) but this trend was not observed in children (10). In addition, a study found that sensitization to rAra h 2 and rAra h 1 and/or rAra h 3 appeared to be predictive of more severe reactions (11). Similarly, the results of a recent study in Australia found that polysensitization to Ara 1, 2 and 3 can help to predict the severity of reaction at challenge (12).
Cross-reactive Molecules
The 11S globulin, Sin a 2, is associated with severe reactions in mustard allergic patients and has a role in cross-reactivity with 11S globulins found in tree nuts and peanuts (6).
Molecular Aspects
Biochemistry
Ara h 3 (glycinin) is a cupin allergen belonging to the legumin family (6, 13). Legumins, also known as 11S globulins, are hexameric proteins present in the seeds of many plants (6). Subunits are synthesized as a single polypeptide which is cleaved to provide an acidic and a basic polypeptide chain, linked by a disulfide bond (6). Ara h 3 has a molecular weight of 60 kDa (14).
Cupins have considerable ability to withstand heat treatment and enzymatic activity (15). Ara h 3 has a role as a storage protein in peanuts (15).
Isoforms, epitopes, antibodies
Two isoforms of Ara h 3, Ara h 3.0101 and Ara h 3.0201, and four IgE-binding epitopes have been identified (14, 16).
Research by Piersma et al. (2005) found that the acidic subunit of Ara h 3 is truncated in several places during processing, which introduces variation. The authors suggested including IgE-binding studies with peanut-derived Ara h 3 when determining the allergenicity of Ara h 3 to reflect the high degree of variation in the Ara h 3 protein structure because this is what peanut-allergic individuals encounter when consuming peanuts (17). 
Cross-reactivity
Frequent cross-reactivity is demonstrated between Ara h 3 and the other cupin allergen found in peanut, Ara h 1, with monosensitization to Ara h 1 and/or Ara h 3 being very uncommon (8).
Cross-reactivity between different legumes is due to the considerable homologies of seed storage proteins, with Ara h 3 having equivalent 11S legumin-like globulins in soya bean (Gly m glycin 1, 2 and 4), pea (Pis s 2), lupin (α-Conglutin) and the condiment, fenugreek (“Tri f 3”) (15). It has been reported that the soybean glycinin exhibits high sequence identity of 62% with peanut glycinin, Ara h 3 (18).
IgE-binding epitopes of Ara h 3 demonstrated structural homology among legumin peanut and tree nut allergens, specifically Jug r 4 of walnut, Cor a 9 of hazelnut and Ana o 2 of cashew nut, helping to explain the observed IgE-binding cross-reactivity (19).
Sin a 2, a major allergen of yellow mustard seed, was shown to share sequence identity with other allergenic 11S globulins, 27% for Ara h 3 and in addition, three epitopes in Ara h 3 were moderately conserved in Sin a 2 which could have an impact in terms of cross-reactivity (16).
Diagnostic Relevance
Disease Severity
The authors who published results from a cross-sectional study involving 222 children in Australia reported that using a combination of Ara h 1, 2 and 3 for peanut component testing could be helpful to identify patients with peanut allergy (12). An earlier study found that using the combination of Ara h 1, 2, and 3 resulted in a higher specificity (94%) when diagnosing peanut allergy in Japanese children in comparison to using IgE to Ara h 2 alone resulting in a sensitivity and specificity of 88% and 84%, respectively. However, the sensitivity of using the combination of Ara h 1, 2, and 3 was 31% (20). A study found that cosensitization to rAra h 2 and rAra h 1 and/or rAra h 3 was predictive of more severe reactions in peanut allergy (11).
Cross-Reactivity
Often, specific IgE measurements for Ara h 1 and Ara h 3 are not necessary due to the high level of cross-reactivity between them, and monosensitization to these seed storage allergen components is uncommon (8).
Compiled By
Author: RubyDuke Communications
Reviewer: Dr. Magnus Borres
Last reviewed: December 2020
References
  1. Lieberman JA, Gupta R, Knibb RC, Haselkorn T, Tilles S, Mack DP, et al. The Global Burden of Illness of Peanut Allergy: A Comprehensive Literature Review. Allergy. 2021;76(5):1367–84.
  2. Sicherer SH. Clinical update on peanut allergy. Ann Allergy Asthma Immunol. 2002;88(4):350-61; quiz 61-2, 94.
  3. Kleber-Janke T, Crameri R, Appenzeller U, Schlaak M, Becker WM. Selective Cloning of Peanut Allergens, Including Profilin and 2S Albumins, by Phage Display Technology. International Archives of Allergy and Immunology. 1999;119(4):265-74.
  4. Hemmings O, Du Toit G, Radulovic S, Lack G, Santos AF. Ara h 2 is the dominant peanut allergen despite similarities with Ara h 6. J Allergy Clin Immunol. 2020;146(3):621-30.e5.
  5. Valcour A, Jones JE, Lidholm J, Borres MP, Hamilton RG. Sensitization profiles to peanut allergens across the United States. Annals of Allergy, Asthma & Immunology. 2017;119(3):262-6.e1.
  6. Bublin M, Breiteneder H. Cross-Reactivity of Peanut Allergens. Current Allergy and Asthma Reports. 2014;14(4):426.
  7. Chassaigne H, Nørgaard JV, Hengel AJ. Proteomics-based approach to detect and identify major allergens in processed peanuts by capillary LC-Q-TOF (MS/MS). J Agric Food Chem. 2007;55(11):4461-73.
  8. Matricardi PM, Kleine-Tebbe J, Hoffmann HJ, Valenta R, Hilger C, Hofmaier S, et al. EAACI Molecular Allergology User's Guide. Pediatr Allergy Immunol. 2016;27 Suppl 23:1-250.
  9. Johnson J, Malinovschi A, Lidholm J, Petersson CJ, Nordvall L, Janson C, et al. Sensitization to storage proteins in peanut and hazelnut is associated with higher levels of inflammatory markers in asthma. Clin Mol Allergy. 2020;18:11.
  10. Klemans RJ, Liu X, Knulst AC, Knol MJ, Gmelig-Meyling F, Borst E, et al. IgE binding to peanut components by four different techniques: Ara h 2 is the most relevant in peanut allergic children and adults. Clin Exp Allergy. 2013;43(8):967-74.
  11. Astier C, Morisset M, Roitel O, Codreanu F, Jacquenet S, Franck P, et al. Predictive value of skin prick tests using recombinant allergens for diagnosis of peanut allergy. J Allergy Clin Immunol. 2006;118(1):250-6.
  12. Kaur N, Mehr S, Katelaris C, Wainstein B, Altavilla B, Saad R, et al. Added Diagnostic Value of Peanut Component Testing: A Cross-Sectional Study in Australian Children. J Allergy Clin Immunol Pract. 2021;9(1):245–253.
  13. Koppelman SJ, Knol EF, Vlooswijk RA, Wensing M, Knulst AC, Hefle SL, et al. Peanut allergen Ara h 3: isolation from peanuts and biochemical characterization. Allergy. 2003;58(11):1144-51.
  14. IUIS. WHO/IUIS Allergen Database 2020 [cited 2020 November]. Available from: http://www.allergen.org/.
  15. Fæste CK, Namork E. Differentiated Patterns of Legume Sensitisation in Peanut-Allergic Patients. Food Analytical Methods. 2010;3(4):357-62.
  16. Palomares O, Vereda A, Cuesta-Herranz J, Villalba M, Rodríguez R. Cloning, sequencing, and recombinant production of Sin a 2, an allergenic 11S globulin from yellow mustard seeds. J Allergy Clin Immunol. 2007;119(5):1189-96.
  17. Piersma SR, Gaspari M, Hefle SL, Koppelman SJ. Proteolytic processing of the peanut allergen Ara h 3. Mol Nutr Food Res. 2005;49(8):744-55.
  18. Beardslee TA, Zeece MG, Sarath G, Markwell JP. Soybean glycinin G1 acidic chain shares IgE epitopes with peanut allergen Ara h 3. Int Arch Allergy Immunol. 2000;123(4):299-307.
  19. Barre A, Jacquet G, Sordet C, Culerrier R, Rougé P. Homology modelling and conformational analysis of IgE-binding epitopes of Ara h 3 and other legumin allergens with a cupin fold from tree nuts. Mol Immunol. 2007;44(12):3243-55.
  20. Ebisawa M, Movérare R, Sato S, Maruyama N, Borres MP, Komata T. Measurement of Ara h 1-, 2-, and 3-specific IgE antibodies is useful in diagnosis of peanut allergy in Japanese children. Pediatr Allergy Immunol. 2012;23(6):573-81.