Recent searches Clear History
Kontaktieren Sie uns
Allergen Encyclopedia
Table of Contents
Komponente
f76 nBos d 4 α-lactalbumin, Milk
f76 nBos d 4 α-lactalbumin, Milk Scientific Information
Typ:
Component
Name; WHO/IUIS:
nBos d 4 α-lactalbumin, Milk
Allergen code:
f76
Source Material:
nBos d 4 is purified from milk extract
Allergen
Cow's milk is popularly consumed throughout the world. Allergy to cow's milk is prevalent among infants and young children during the first year of life, affecting 0.5-3% at the age of 1 year. Bos d 4 (α-lactalbumin) has been reported as a major allergenic protein found in cow's milk whey fraction. The prevalence of sensitization to Bos d 4 among cow’s milk allergy (CMA) patients varies up to 80%. Milk whey proteins like Bos d 4 are heat-labile and therefore are likely to be destroyed after extensive cooking and baking of milk. Thus, specific immunoglobulin E to Bos d 4 might help assess tolerance to cooked or baked milk. Clinical manifestations of Bos d 4-mediated allergy majorly include anaphylaxis and atopic dermatitis. Cross-reactivity has been observed between antibodies against bovine Bos d 5 (β-lactoglobulin) and Bos d 4 in cow's milk due to the shared amino acid sequence. Bos d 4, present in cow and human milk, was also found to be cross-reactive. Moreover, Bos d 4 from cow's milk has been found to exhibit sequence homology with other mammalian milk proteins (like water buffalo, ewe, goat, pig, donkey, horse, and camel).
Epidemiology
Worldwide distribution
Cow’s milk is popularly consumed worldwide by infants, children, and adults (Muthukumar, Selvasekaran et al. 2020). Allergy to cow’s milk is prevalent among infants and young children, with a high prevalence observed during the first year of life (Kaczmarski, Wasilewska et al. 2013, Vandenplas 2017), affecting 0.5-3% at the age of 1 year (Skripak, Matsui et al. 2007, Flom and Sicherer 2019).
Bos d 4 (α-lactalbumin; ALA), Bos d 5 (β-lactoglobulin, BLG) and Bos d 8 (casein) are considered major allergens present in cow’s milk (Villa, Costa et al. 2018). According to World Allergy Organization (WAO) Diagnosis and Rationale for Action against Cow’s Milk Allergy (DRACMA) Guidelines, the prevalence of allergic reactions towards Bos d 4 among cow’s milk allergy (CMA) patients ranges between 0 and 80% (Fiocchi, Brozek et al. 2010).
A Taiwan-based study reported sensitization towards Bos d 4 among 60% (114/190) of atopic individuals with specific immunoglobulin E (sIgE) towards whole cow’s milk. Among all three milk allergens (Bos d 4, Bos d 5 and Bos d 8) tested, the sensitization towards Bos d 4 and the mean serum level of Bos d 4 sIgEs were found to be the highest (Chen, Lee et al. 2014). Furthermore, a Poland-based study conducted among 291 children (2-14 years) with a history of CMA also reported sensitization towards Bos d 4 in 18.8% (13/69) of children (Kaczmarski, Wasilewska et al. 2013).
A study in Iran reported allergy against Bos d 4 in 78.3% of patients with CMA (n=86) (Shokouhi Shoormasti, Fazlollahi et al. 2016). Furthermore, a study in Italy conducted on 104 children with suspected CMA (n=58) found the prevalence of IgE reactivity towards Bos d 4 to be 27.6% (16/58) (D'Urbano, Pellegrino et al. 2010).Bos d 4 has been reported as a major allergenic protein in cow’s milk, and 51% of patients (n=92) allergic to cow’s milk have reported sensitization toward Bos d 4 (Wal 2002).
Environmental Characteristics
Source and tissue
Cow’s milk contains mainly two types of proteins, casein (Bos d 8; constitutes 80% of the total milk protein) and whey (such as Bos d 4 and Bos d 5; comprises approximately 20% of the total milk protein) (Flom and Sicherer 2019).
Bos d 4 is synthesized in the mammary gland, expressed exclusively during lactation, and is one of the most commonly found proteins in most mammalian milk (Wal 2002, Fiocchi, Brozek et al. 2010, Azdad, Mejrhit et al. 2018).
Bos d 4 belongs to the lysozyme superfamily of protein and is a regulatory subunit of lactose synthase. It alters the substrate specificity of the enzyme galactosyltransferase in the mammary gland and aids in the biosynthesis of lactose via interaction with lactose synthase (Fiocchi, Brozek et al. 2010).
According to the World Health Organization (WHO) and International Union of Immunological Societies (IUIS) database, Bos d 4 (ALA) has been identified and listed as a food allergen from cow’s milk (WHO/IUIS 2020).
Clinical Relevance
Disease severity
Cow milk allergy is considered one of the most common pediatric food allergies (Agyemang, Saf et al. 2019). Milk allergy can be either IgE-mediated or non-IgE-mediated (Walsh, Meyer et al. 2016). The clinical manifestations of cow milk protein allergy are reported to be of two types such as immediate (shortly after consumption) and delayed (after hours or days of consumption) responses (De Greef, Hauser et al. 2012). The immediate reactions are generally IgE-mediated and can lead to angioedema, urticaria, aggravation of atopic dermatitis (AD), vomiting, in some cases, anaphylaxis, as well as respiratory symptoms (breathing, wheezing, coughing) (De Greef, Hauser et al. 2012, Ito, Futamura et al. 2012, Walsh, Meyer et al. 2016, Flom and Sicherer 2019). On the other hand, most delayed reactions are non-IgE mediated (Walsh, Meyer et al. 2016) and can lead to gastrointestinal symptoms (like enteropathy or proctocolitis) or skin reactions (such as AD) (De Greef, Hauser et al. 2012, Ito, Futamura et al. 2012).
Milk whey proteins (such as Bos d 4) are heat-labile and therefore destroyed by extensive cooking and baking of milk (Wal 2002). Thus, the sIgE level to Bos d 4 may help assess the likelihood of tolerance to cooked or baked milk, and 75% of children with CMA may actually tolerate baked milk products such as muffins, waffles, cakes, and bread (Nowak-Wegrzyn, Bloom et al. 2008).
According to a study, anaphylaxis was reported in 6/15 patients after ingestion of cow’s milk. Furthermore, sIgE against Bos d 4 (18.8 ± 19.6 kU/L (median: 16, range: 0.51–54.9) was observed in these patients (Kiykim, Karakoc-Aydiner et al. 2019).
A study conducted among 950 children with clinically diagnosed AD reported that 60.1% (283/471) of children affected with AD and sensitized to cow’s milk (n=471) exhibited positive sIgE towards Bos d 4 (Lee, Yoon et al. 2013).
Molecular Aspects
Biochemistry
Bos d 4 is a globular protein (monomeric) of molecular weight 14.4 kDa, consisting of 123 amino acids. The protein contains four disulfide bonds within its molecule, which provide stability to its structure (Wal 2002, Tsabouri, Douros et al. 2014). It thereby has a stable protein structure with the ability to refold. The secondary structure of the protein is highly ordered and is stabilized by the presence of a calcium-binding high-affinity site (Tsabouri, Douros et al. 2014, Villa, Costa et al. 2018). The tertiary structure of this protein is spherical and rigid with an “elbow-Ca2+-binding loop” and 2 other structural domains, including an α-helical domain and a short β-sheet domain at N-terminal and C-terminal, respectively. The β-sheet domain is adjacent to the calcium-binding loop (Villa, Costa et al. 2018) and comprises 79-88 amino acid residues (Hochwallner, Schulmeister et al. 2014). In addition, cysteine residues are present on both sides of the loop, which creates four disulfide bridges, thus closing the loop and helping to stabilize the Bos d 4 structure (Hochwallner, Schulmeister et al. 2014). This protein is responsible for regulating the enzyme system of galactosyltransferase, which is involved in the biosynthesis of lactose (Wal 2002, Tsabouri, Douros et al. 2014).
Isoforms, epitopes, antibodies
According to a study, the conformation of the allergenic loops in Bos d 4 and Bos d 5 are found to have similarities, indicating the presence of characteristic conformation in their allergenic sites. The loops are distinct and firmly held as strings at both ends by antiparallel strands (Sharma, Kumar et al. 2001). The structure of sequential epitopes recognized by IgE antibodies to Bos d 4 indicated the presence of 4 IgE-binding regions (El-Agamy 2007). Likewise, in another study on 11 persistent CMA patients (4-18 years), 4 IgE-binding and 3 immunoglobulin G binding sites on Bos d 4 were identified. The IgE binding epitopes on Bos d 4 are mostly conformational rather than sequential (Järvinen, Chatchatee et al. 2001).
The epitopes are mostly present at the N- and C-terminals of the protein. The IgE epitopes remain in clusters, which may lead to cross-linking of IgE antibodies efficiently on effector cells and affect the protein's allergenic potential (Hochwallner, Schulmeister et al. 2014).
Both Bos d 4 and Bos d 5 are heat-sensitive proteins, but Bos d 4 is found to be more stable than Bos d 5 based on the molecular structure. The antigenic potential of Bos d 4 has been reported to enhance progressively with the rise in temperature from 50o to 90oC; however, a sharp reduction occurs with a further increase in temperature from 90o to 120oC. Moreover, it has been observed that heating of Bos d 4 at 120oC for 20 min lowers its allergenicity below the baseline value (Bu, Luo et al. 2009). A Moroccan study of 557 CMA patients revealed an average reduction of the allergenic potential of Bos d 4 (lowering of IgE binding) on heating at 90oC and pepsin treatment at 37oC for 1 hour by 59% and 74%, respectively (Azdad, Mejrhit et al. 2018).
Bos d 4 and Bos d 5 are reported to be heat-labile (destabilization of conformational epitopes occurs due to extensive heating). Moreover, clinical implications of Bos d 4 and Bos d 5 have been reported in patients; thus, patients reactive towards only Bos d 4 and/or Bos d 5 might be able to tolerate foods containing milk that has been extensively cooked or baked (Nowak-Wegrzyn, Bloom et al. 2008, Villa, Costa et al. 2018).
To date (7th June 2022), only one isoallergen of Bos d 4, i.e., Bos d 4.0101, has been identified and officially published by the WHO/IUIS Allergen Nomenclature Subcommittee (WHO/IUIS 2020).
Cross-reactivity
Milk of different ruminant species (such as cow, goat, and sheep) is composed of homologous proteins with similar structures and functions, thereby leading to cross-reactivity between the milk proteins (Villa, Costa et al. 2018).
A prospective cohort study conducted on 38 children (age <14 years) with CMA-associated symptoms detected the rate of cross-sensitization (skin-prick test-based) between goat milk and cow milk to be 63.2% (24/38) (Ehlayel, Bener et al. 2011).
A study reported that IgEs present in the sera of CMA children are able to recognize other mammalian (goat, sheep, and buffalo) milk proteins. However, cross-reactivity was found to be weak with milk proteins of donkeys and mares, and almost none with camel milk protein (Restani, Gaiaschi et al. 1999, El-Agamy 2007).
A study conducted among 20 patients with CMA demonstrated cross-reactivity between Bos d 4 present in cow and human milk. The results signified a greater amino acid sequence similarity between these two proteins (Maynard, Chatel et al. 1999).
Antibodies against bovine Bos d 5 have been found to exhibit cross-reactivity with the Bos d 4 present in cow’s milk. This cross-reactivity is reported due to the shared amino acid sequence between these two proteins (Baroglio, Giuffrida et al. 1998).
Bos d 4 from cow’s milk protein shows sequence homology with water buffalo (99.3%), ewe (97.2%), goat (95.1%), pig (74.6%), donkey (71.5%), horse (72.4%), and camel (69.7%) (Tsabouri, Douros et al. 2014).
Diagnostic Relevance
Exposure
Cow milk allergy mainly occurs through ingestion (Kaczmarski, Wasilewska et al. 2013).
Compiled By
Author: Turacoz Healthcare Solutions
Reviewer: Dr. Eva Södergren
Explained Results
Allergen Information
Bos d 4 (α-lactalbumin) is considered a major allergenic protein in cow’s milk (Villa, Costa et al. 2018).
Clinical relevance
Typical symptoms of CMA include skin reactions (angioedema, urticaria, and atopic dermatitis) (De Greef, Hauser et al. 2012, Ito, Futamura et al. 2012, Lee, Yoon et al. 2013, Flom and Sicherer 2019), gastrointestinal symptoms (De Greef, Hauser et al. 2012), respiratory systems, and in some cases, anaphylaxis (Kiykim, Karakoc-Aydiner et al. 2019).
Cross-reactivity
Antibodies against bovine Bos d 4 show cross-reactivity with Bos d 5 present in cow’s milk due to shared amino acid sequence (Baroglio, Giuffrida et al. 1998). Bos d 4 from cow’s milk shows sequence homology with other mammalian milk proteins, including water buffalo, ewe, goat, pig, donkey, horse, and camel (Tsabouri, Douros et al. 2014). Furthermore, cross-reactivity has also been reported between Bos d 4 in cow and human milk (Maynard, Chatel et al. 1999).
References
  1. Agyemang, A., S. Saf, T. Sifers, M. Mishoe, M. P. Borres, H. A. Sampson and A. Nowak-Wegrzyn (2019). "Utilizing boiled milk sIgE as a predictor of baked milk tolerance in cow's milk allergic children." J Allergy Clin Immunol Pract 7(6): 2049-2051.
  2. Azdad, O., N. Mejrhit and L. Aarab (2018). "Reduction of the allergenicity of cow's milk alpha-lactalbumin under heat-treatment and enzymatic hydrolysis in Moroccan population." Eur Ann Allergy Clin Immunol 50(4): 177-183.
  3. Baroglio, C., M. G. Giuffrida, A. Cantisani, L. Napolitano, E. Bertino, C. Fabris and A. Conti (1998). "Evidence for a common epitope between bovine alpha-lactalbumin and beta-lactoglobulin." Biol Chem 379(12): 1453-1456.
  4. Bu, G., Y. Luo, Z. Zheng and H. Zheng (2009). "Effect of heat treatment on the antigenicity of bovine α-lactalbumin and β-lactoglobulin in whey protein isolate." Food and Agricultural Immunology 20(3): 195-206.
  5. Chen, F. M., J. H. Lee, Y. H. Yang, Y. T. Lin, L. C. Wang, H. H. Yu and B. L. Chiang (2014). "Analysis of alpha-lactalbumin-, beta-lactoglobulin-, and casein-specific IgE among children with atopic diseases in a tertiary medical center in Northern Taiwan." J Microbiol Immunol Infect 47(2): 130-136.
  6. D'Urbano, L. E., K. Pellegrino, M. C. Artesani, S. Donnanno, R. Luciano, C. Riccardi, A. E. Tozzi, L. Rava, F. De Benedetti and G. Cavagni (2010). "Performance of a component-based allergen-microarray in the diagnosis of cow's milk and hen's egg allergy." Clin Exp Allergy 40(10): 1561-1570.
  7. De Greef, E., B. Hauser, T. Devreker, G. Veereman-Wauters and Y. Vandenplas (2012). "Diagnosis and management of cow’s milk protein allergy in infants." World Journal of Pediatrics 8(1): 19-24.
  8. Ehlayel, M., A. Bener, K. Abu Hazeima and F. Al-Mesaifri (2011). "Camel milk is a safer choice than goat milk for feeding children with cow milk allergy." ISRN Allergy 2011: 391641.
  9. El-Agamy, E. (2007). "The challenge of cow milk protein allergy." Small Ruminant Research 68(1-2): 64-72.
  10. Fiocchi, A., J. Brozek, H. Schünemann, S. L. Bahna, A. von Berg, K. Beyer, M. Bozzola, J. Bradsher, E. Compalati, M. Ebisawa, M. A. Guzman, H. Li, R. G. Heine, P. Keith, G. Lack, M. Landi, A. Martelli, F. Rancé, H. Sampson, A. Stein, L. Terracciano and S. Vieths (2010). "World Allergy Organization (WAO) Diagnosis and Rationale for Action against Cow's Milk Allergy (DRACMA) Guidelines." The World Allergy Organization journal 3(4): 57-161.
  11. Flom, J. D. and S. H. Sicherer (2019). "Epidemiology of Cow's Milk Allergy." Nutrients 11(5).
  12. Hochwallner, H., U. Schulmeister, I. Swoboda, S. Spitzauer and R. Valenta (2014). "Cow's milk allergy: from allergens to new forms of diagnosis, therapy and prevention." Methods 66(1): 22-33.
  13. Ito, K., M. Futamura, R. Moverare, A. Tanaka, T. Kawabe, T. Sakamoto and M. P. Borres (2012). "The usefulness of casein-specific IgE and IgG4 antibodies in cow's milk allergic children." Clin Mol Allergy 10(1): 1.
  14. Järvinen, K. M., P. Chatchatee, L. Bardina, K. Beyer and H. A. Sampson (2001). "IgE and IgG binding epitopes on alpha-lactalbumin and beta-lactoglobulin in cow's milk allergy." Int Arch Allergy Immunol 126(2): 111-118.
  15. Kaczmarski, M., J. Wasilewska, B. Cudowska, J. Semeniuk, M. Klukowski and E. Matuszewska (2013). "The natural history of cow's milk allergy in north-eastern Poland." Adv Med Sci 58(1): 22-30.
  16. Kiykim, A., E. Karakoc-Aydiner, E. Gunes, E. Nain, I. Ogulur, D. Yazici, S. Aktac, A. H. Bicer, C. Sackesen, S. Baris and A. Ozen (2019). "Evaluation of a Standardized Bakery Product (SUTMEK) as a Potential Tool for Baked-Milk Tolerance and Immunotherapy Research Studies." Int Arch Allergy Immunol 178(1): 1-9.
  17. Lee, J. M., J. S. Yoon, S. A. Jeon and S. Y. Lee (2013). "Sensitization patterns of cow's milk and major components in young children with atopic dermatitis." Asia Pac Allergy 3(3): 179-185.
  18. Maynard, F., J.-M. Chatel and J.-M. Wal (1999). "Immunological IgE Cross-Reactions of Bovine and Human α-Lactalbumins in Cow's Milk Allergic Patients." Food and Agricultural Immunology 11(2): 179-189.
  19. Muthukumar, J., P. Selvasekaran, M. Lokanadham and R. Chidambaram (2020). "Food and food products associated with food allergy and food intolerance - An overview." Food Res Int 138(Pt B): 109780.
  20. Nowak-Wegrzyn, A., K. A. Bloom, S. H. Sicherer, W. G. Shreffler, S. Noone, N. Wanich and H. A. Sampson (2008). "Tolerance to extensively heated milk in children with cow's milk allergy." J Allergy Clin Immunol 122(2): 342-347, 347 e341-342.
  21. Restani, P., A. Gaiaschi, A. Plebani, B. Beretta, G. Cavagni, A. Fiocchi, C. Poiesi, T. Velona, A. G. Ugazio and C. L. Galli (1999). "Cross-reactivity between milk proteins from different animal species." Clin Exp Allergy 29(7): 997-1004.
  22. Sharma, S., P. Kumar, C. Betzel and T. P. Singh (2001). "Structure and function of proteins involved in milk allergies." Journal of Chromatography B: Biomedical Sciences and Applications 756(1): 183-187.
  23. Shokouhi Shoormasti, R., M. R. Fazlollahi, S. Barzegar, P. Teymourpour, Z. Yazdanyar, Z. Lebaschi, M. Nourizadeh, B. Tazesh, M. Movahedi, H. Kashani, Z. Pourpak and M. Moin (2016). "The Most Common Cow's Milk Allergenic Proteins with Respect to Allergic Symptoms in Iranian Patients." Iran J Allergy Asthma Immunol 15(2): 161-165.
  24. Skripak, J. M., E. C. Matsui, K. Mudd and R. A. Wood (2007). "The natural history of IgE-mediated cow's milk allergy." J Allergy Clin Immunol 120(5): 1172-1177.
  25. Tsabouri, S., K. Douros and K. N. Priftis (2014). "Cow's milk allergenicity." Endocr Metab Immune Disord Drug Targets 14(1): 16-26.
  26. Vandenplas, Y. (2017). "Prevention and Management of Cow's Milk Allergy in Non-Exclusively Breastfed Infants." Nutrients 9(7).
  27. Villa, C., J. Costa, M. B. P. P. Oliveira and I. Mafra (2018). "Bovine Milk Allergens: A Comprehensive Review." Comprehensive Reviews in Food Science and Food Safety 17(1): 137-164.
  28. Wal, J.-M. (2002). "Cow's milk proteins/allergens." Annals of Allergy, Asthma & Immunology 89(6, Supplement): 3-10.
  29. Walsh, J., R. Meyer, N. Shah, J. Quekett and A. T. Fox (2016). "Differentiating milk allergy (IgE and non-IgE mediated) from lactose intolerance: understanding the underlying mechanisms and presentations." Br J Gen Pract 66(649): e609-611.
  30. WHO/IUIS. (2020). "Bos d 4."   Retrieved 07-06-2022, from http://www.allergen.org/viewallergen.php?aid=162.