Recent searches Clear History
Kontaktieren Sie uns
Allergen Encyclopedia
Table of Contents
Komponente
o215 Alpha-Gal
o215 Alpha-Gal Scientific Information
Typ:
Component
Name; WHO/IUIS:
Alpha-Gal
Route of Exposure:
Oral
Biological function:
alpha-Gal is a carbohydrate present on glycoproteins in non-primate mammals and certain invertebrates, but not in humans, Old World monkeys or apes.
Allergen code:
o215
Source Material:
Red meat
Other Names :
GAL (alpha-Gal) Thyroglobulin, bovine,galactose-α-1,3-galactose
Summary
α-Gal-associated meat allergy is a recently described syndrome in which individuals who have been bitten by ticks of the Ixodidae family become sensitized to the carbohydrate determinant galactose-α-1,3-galactose (α-Gal). The resulting IgE antibodies react with α-Gal epitopes on mammalian meat, resulting in an allergic reaction and in some cases anaphylaxis after eating meat or gelatin. A distinctive feature of this allergy is the delayed onset of symptoms, which occurs 2-6 hours after eating meat. The delayed symptom onset is thought to reflect the appearance of glycolipid α-Gal moieties, which are believed to be involved in the allergic reaction, in the bloodstream.
Diagnosis of α-Gal-associated meat allergy may not be straightforward, especially in children, as the syndrome can be confused with protein-based meat allergies. While negative skin prick test (SPT) results are unreliable, the syndrome can be confirmed by correlating clinical history with the measurement of IgE antibodies to α-Gal. If α-Gal sIgE results represent >2% of total sIgE, a diagnosis of meat allergy is very likely (1). Serum tryptase testing can help identify people who may have more severe reactions to tick bites (2). Finally, testing for the presence of IgE to α-Gal could identify a risk factor for medical treatment with cetuximab, eating gelatin-containing substances, or using artificial bovine blood in areas where hard body ticks are common, particularly in individuals with a history of a tick bite or demonstrated allergy to mammalian meat or gelatin (1).
Environmental Characteristics
Worldwide distribution
To date, cases of α-Gal-associated meat allergy have been reported in the southeastern (27) northeastern and upper midwestern US (3), Australia (4) Asia (5) and in several European countries, including France (6-8) Spain(9), Germany(10), Switzerland (11), and Sweden (12), Italy (13).
On all continents, the ticks responsible for sensitization to α-Gal belong to the Ixodidae family of hard ticks, with each continent harboring its native species. In the US, bites from the Lone Star tick Amblyomma americanum (Figure 1) are the predominant, if not exclusive, cause of α-Gal sensitization (14). There is a significant correlation between IgE to Lone Star tick bites and antibodies to α-Gal (15). Warming temperatures and the availability of desirable hosts favor the continued range expansion of Lone Star ticks (3), with reported sightings from Maine to Florida, in Wisconsin (16), and as far west as Texas, Oklahoma and Colorado (17). Borrelia burgdorferi, the bacterium that ticks carry causes Lyme disease, does not appear to induce IgE to α-Gal and its bites are not associated with itching (15). In Australia, the suspected tick is Ixodes holocyclus (7, 18), whereas in Europe it is Ixodes ricinus (19) ,and Hematophagous ixodidae in Asia (20).
The prevalence of detectable IgE antibodies to α-Gal is highest in areas where Ixodidae ticks are common. In Sweden, around 10% of 143 healthy blood donors in the greater Stockholm area had IgE antibodies to α-Gal, compared with only 0.7% in the north of the country where tick bites are rare (12). It is, however, unclear exactly what proportion of sensitized individuals experience allergic symptoms after eating mammalian meat. Commins and Platts-Mills estimated that around 10% of individuals with detectable α-Gal antibodies may display red meat allergy (21). The prevalence of IgE to α-Gal among Danish and Spanish adults was estimated at 5.5% and 8.1%, respectively, for levels ≥0.1 kUA/L, and at 1.8% and 2.2%, respectively, for levels ≥0.35 kUA/L (22). Based on a survey of 1,000 Americans, 7.7% reported a history of an anaphylactic reaction, with an estimated 5.1% of that population having “probable anaphylaxis” and 1.6% “very likely” anaphylaxis (23), 218 of the reported cases met the criteria for anaphylaxis, and 85 were associated with a definitive cause; α-Gal was cited in 28/85 (33%) of cases with a known cause, more frequently than other food allergies (24/85 cases, 28%).
Adult onset originally appeared to be a characteristic of α-Gal-associated meat allergy (24). In a series of 29 patients with α-Gal-associated meat allergy in Japan, 21 were over 60 years of age (5). Nevertheless, cases of α-Gal-associated meat allergy have also been observed in children (15, 25). In a case series of 51 children ages 4-17 years, who had possible delayed allergic reactions to mammalian foods, 45 had high serum IgE levels to α-Gal, which correlated with beef IgE levels (15). Questioning revealed a history of symptoms 3-6 hours after eating meat and of tick bites. Among adults and children, and atopic and non-atopic individuals, the levels of α-Gal IgE and the severity of reactions were similar (26)
Blood group phenotype
The B blood group phenotype appears to confer a protective effect against the production of high levels of IgE to α-Gal, with a strong relationship between α-Gal-associated meat allergy and absence of the B blood group phenotype (12, 26, 27)
A study of the relationship between the B blood group phenotype and IgG and IgE responses to α-Gal found that none of the subjects with the B phenotype expressed IgE to α-Gal (28). Among 39 Swedish patients with delayed meat allergy, only two (5%) had blood group B or AB, which is significantly lower than the expected overall prevalence of 18% in the Swedish population (12). The same study reported that in a group of healthy blood donors and patients with Lyme disease with detectable IgE to α-Gal, the levels of IgE against α-Gal were very low among those with the blood group B phenotype (12). The mechanism of this protective effect has not been fully elucidated. However, the structure of the α-Gal epitope is closely related to that of the blood group B antigen, and it appears that the presence of the group B antigen induces tolerance and eliminates most B cell clones that could otherwise interact with tick-derived α-Gal antigens (28).
In addition, Wilson et al (1) have found indications that a-Gal sensitization could be a risk factor for coronary artery disease. This need to be further studied. 
Figure 1. Structure of galactose-a1,3-galactose
Figure 1. Structure of galactose-a1,3-galactose.
Prevention and Therapy
Management of α-Gal-associated meat allergy (29)
The primary management strategy is avoiding mammalian meat, especially fatty cuts.
  • Avoid meats, including beef, pork, lamb, horse, goat, rabbit, squirrel, and venison.
  • Avoid mammalian organs such as liver, intestine, heart, and kidney (especially pork kidney) and other mammalian products including lard, suet, and pork rinds.
  • Gelatin has reportedly caused severe reactions, either as jelly sweets and marshmallows or as intravenous preparations.
Although most patients tolerate dairy products and gelatin, patients who continue to have unexplained symptoms may also be advised to avoid dairy foods and gelatin.
Avoid additional tick bites by staying away from tick-infested areas, wearing protective clothing, or using products containing N, N-diemethyl-meta-toluamide may make some patients less prone to symptomatic reactions.
Molecular Aspects
Biochemistry
The α-Gal epitope (Galα1-3Galβ1-4GlcNAc-R, or Gaα1- 3Galα1-3GlcNAc-R) is a unique carbohydrate structure present in all mammals except humans and Old World monkeys (Figure 2), where it exists on the cell surface as both glycolipids and glycoproteins (30). The onset of clinical symptoms in α-Gal-associated meat allergy corresponds with basophil activation, implying that it coincides with the appearance of the antigen in the bloodstream (31). Lipids enter the bloodstream 3–4 hours after a meal, suggesting that the delay in symptom onset could reflect the time required for absorption and digestion of glycolipid α-Gal antigens (21). The observation that fatty meats provoke more consistent and severe reactions is also consistent with the notion that glycolipids may play an important role (21, 31). This hypothesis is further supported by evidence that glycolipids can elicit a robust immune response in humans (32). However, a recent screen of IgE-binding proteins from beef and pork, using sera from Japanese patients with α-Gal-associated meat allergy, identified α-Gal on the glycoproteins laminin γ-1 and the collagen α 1 (VI) chain as likely common IgE-reactive proteins in these patients with beef allergy (5). 
Figure 2. Levels of a-Gal-specific IgE increase after tick bites and decline with time after the bite. Adapted from (14).
Figure 2. Levels of a-Gal-specific IgE increase after tick bites and decline with time after the bite. Adapted from (14).
Comparison with other meat allergies
Meat allergy
Despite a high level of meat consumption in developed countries, allergy to meat is uncommon (33, 34) it is normally outgrown during the first years of life and is rare in adults (35). Meat allergy has been estimated to occur in around 3% to >7% of children and adults with food allergies (36, 37). Allergy to beef is the most commonly reported form of meat allergy, affecting 1.5 to 6.5% of children with atopic dermatitis or food allergies (35, 38) which corresponds to a prevalence of around 0.3% in the general population (39).
Table 1. Features of α-Gal-mediated meat allergy, protein-mediated meat allergy (40-42), and pork-cat syndrome (43, 44)
α-Gal-mediated meat allergy  
(1, 31)
Protein-mediated meat allergy
 (1, 33, 35, 40-42, 45, 46)
Pork-cat syndrome 
(1, 43, 44)
Sensitizing allergen
α-Gal epitope from a tick bite.
Bovine serum albumin (Bos d 6), or immunoglobulin (Bos d 7), actin, myoglobin
Chicken serum albumin (Gal d 5)
Cat serum albumin (Fel d 2)
Clinical Features
Triggers
α-Gal on mammalian meat and offal proteins, milk in some patients.
 Gelatin from mammalian sources.
Co-factors can increase the sensitivity to food triggers.
Non-oral triggers: Cetuximab infusion, vaccines, and colloids containing mammalian gelatin
Sensitizing meat proteins
Pork serum albumin
Symptoms
Often severe, ranging from generalized pruritus and urticaria, to gastrointestinal (GI) symptoms, angioedema, and anaphylaxis.
Typically less severe, but range from conjunctivitis, urticaria, angioedema, and dyspnea to anaphylaxis.
Typically less severe, but ranging from abdominal cramping, diarrhea, nausea, itching, hives to anaphylaxis.
Oral pruritus can occur during the meal.
Symptom onset
Delayed reactions – generally 3-6 hours after consuming mammalian meat, but longer delays are not uncommon.
Co-factors can reduce time to onset.
Rapid - within 2 hours of consuming meat.  Rapid – usually within 1 hour of consuming pork.
Age group affected
Predominantly adults, but also children.
Predominantly children.
Predominantly adults and teenagers.
Diagnosis
Skin prick testing
Unreliable - many patients show negative SPT results with meat extracts and fresh meat preparations. Positive to mammalian serum albumins and beef, lamb, pork, and rabbit meat (can be negative to these meats after, cooking) Positive to cat and other animal danders and meats.
Immunoassay
Can show positive results to beef, pork, milk, cat, and dog dander.
Can be positive to mammalian meats (beef, lamb, pork, rabbit), serum albumins and epithelium (dog, cat, cow, sheep, pig), and to cow’s milk. 
Can be positive for pork and beef, and mammalian serum albumin (cat, dog, cow).
Patients with meat allergy are often sensitized to cross-reactive proteins, such as serum albumin, gamma globulins, tropomyosins, and actin (40). Consequently, many patients show cross-reactivity to animal danders in SPT and immunoassays (40, 47) and a high proportion of beef-allergic patients also show clinical cross-reactions to cow’s milk (35). In contrast with α-Gal-associated meat allergy, symptoms in individuals sensitized to protein epitopes usually appear immediately after meat is eaten (41, 42) (Table 1). 
Diagnostic Relevance
Disease Severity
Delayed onset of symptoms is the most clinically distinguishing characteristic of α-Gal-associated meat allergy compared with protein-based meat allergies. However, accurate information on the timing of symptom onset is not always available from the patient.
Exposure
Alpha-Gal is present in all mammals except for humans and old-world monkeys (30). Therefore, red meats such as beef, pork, and lamb are the most common triggers. But derivatives of red meat can be hiding in a variety of foods that aren’t always clearly labeled, like gelatin-containing foods and dairy. These derivatives can also be found in many medications and biologic therapies of which patients need to be aware(1, 4).
Not all triggers are equally likely to cause a reaction—see the chart below for triggers ranked from higher to lower risk. 
Table 2.  Adapted from (1) 
Food Medications/Biologic Therapies
Beef, pork, lamb, innards
Dairy
Gelatin-containing foods
Cetuximab
Gelatin plasma expanders
Anti-venom (e.g., CroFab)
Bovine/porcine heart valves
Gelatin-containing vaccines (e.g., Zostavax, MMR)
Pancreatic enzyme replacement (e.g., pancrelipase)
Heparin
Gelcaps
There are also cofactors that may increase the risk or severity of the reaction, including:
  • Non-steroidal anti-inflammatory drugs
  • Exercise
  • Alcohol
  • Recent tick bites (1, 45)
Compiled By
Reviewer: Dr. Christian Fischer
Last reviewed: October 2020
References
  1. Platts-Mills TAE, Li R-C, Keshavarz B, Smith AR, Wilson JM. Diagnosis and Management of Patients with the α-Gal Syndrome. The Journal of Allergy and Clinical Immunology: In Practice. 2020;8(1):15-23.e1.
  2. 2020. ASoCIaATawaoaA.
  3. Molaei G, Little EAH, Williams SC, Stafford KC. Bracing for the Worst — Range Expansion of the Lone Star Tick in the Northeastern United States. New England Journal of Medicine. 2019;381(23):2189-92.
  4. Mullins RJ, James H, Platts-Mills TAE, Commins S. Relationship between red meat allergy and sensitization to gelatin and galactose-α-1,3-galactose. J Allergy Clin Immunol. 2012;129(5):1334-42.e1.
  5. Takahashi H, Chinuki Y, Tanaka A, Morita E. Laminin γ-1 and collagen α-1 (VI) chain are galactose-α-1,3-galactose-bound allergens in beef. Allergy. 2014;69(2):199-207.
  6. Renaudin J, Jacquenet S, Metz-Favre C, Baudouin E, Engel F, De Blay F, et al. Interest Of Specific Ige Measurement For Galactose-alpha-1,3-galactose In Unexplained Recurrent Urticaria With Angioedema, Predominantly Nocturnal: About 6 Cases. Journal of Allergy and Clinical Immunology. 2012;129(2):AB177.
  7. Morisset M, Richard C, Astier C, Jacquenet S, Croizier A, Beaudouin E, et al. Anaphylaxis to pork kidney is related to IgE antibodies specific for galactose-alpha-1,3-galactose. Allergy. 2012;67(5):699-704.
  8. Jacquenet S, Moneret-Vautrin D-A, Bihain BE. Mammalian meat–induced anaphylaxis: Clinical relevance of anti–galactose-α-1,3-galactose IgE confirmed by means of skin tests to cetuximab. 2009;124(3):603-5.
  9. Nuñez R, Carballada F, Gonzalez-Quintela A, Gomez-Rial J, Boquete M, Vidal C. Delayed mammalian meat–induced anaphylaxis due to galactose-α-1,3-galactose in 5 European patients. Journal of Allergy and Clinical Immunology. 2011;128(5):1122-4.e1.
  10. Jappe U. [Update on meat allergy. α-Gal: a new epitope, a new entity?]. Der Hautarzt; Zeitschrift fur Dermatologie, Venerologie, und verwandte Gebiete. 2012;63(4):299-306.
  11. Michel S, Scherer K, Heijnen IA, Bircher AJ. Skin prick test and basophil reactivity to cetuximab in patients with IgE to alpha-gal and allergy to red meat. Allergy. 2014;69(3):403-5.
  12. Hamsten C, Tran TAT, Starkhammar M, Brauner A, Commins SP, Platts-Mills TAE, et al. Red meat allergy in Sweden: Association with tick sensitization and B-negative blood groups. Journal of Allergy and Clinical Immunology. 2013;132(6):1431-4.e6.
  13. Uasuf CG, Torina A, Ferrantelli V, Brusca I. An unusual case of positive sIgE to Galactose-alpha-1,3-galactose from South Italy. European Annals of Allergy and Clinical Immunology. 2018;50(01):45.
  14. Commins SP, James HR, Kelly LA, Pochan SL, Workman LJ, Perzanowski MS, et al. The relevance of tick bites to the production of IgE antibodies to the mammalian oligosaccharide galactose-α-1,3-galactose. Journal of Allergy and Clinical Immunology. 2011;127(5):1286-93.e6.
  15. Steinke JW, Platts-Mills TAE, Commins SP. The alpha-gal story: Lessons learned from connecting the dots. Journal of Allergy and Clinical Immunology. 2015;135(3):589-96.
  16. Balzer D. Infectious diseases A–Z: Lone star tick found in Wisconsin ltmaJ, 2019. https://newsnetwork.mayoclinic.org/discussion/infectious-diseases-a-z-lone-star-tick-found-in-wisconsin-linked-to-meat-allergy/. .
  17. https://coloradoticks.org/the-ticks/lone-star-tick/. CT-BDAALst.
  18. Van Nunen SA, O’Connor KS, Clarke LR, Boyle RX, Fernando SL. An association between tick bite reactions and red meat allergy in humans. Medical Journal of Australia. 2009;190(9):510-1.
  19. Hamsten C, Starkhammar M, Tran TA, Johansson M, Bengtsson U, Ahlén G, et al. Identification of galactose-α-1,3-galactose in the gastrointestinal tract of the tick Ixodes ricinus; possible relationship with red meat allergy. Allergy. 2013;68(4):549-52.
  20. Wen L, Zhou J, Yin J, Sun J-L, Sun Y, Wu K, et al. Delayed Anaphylaxis to Red Meat Associated With Specific IgE Antibodies to Galactose. Allergy, Asthma & Immunology Research. 2015;7(1):92.
  21. Commins SP, Platts-Mills TAE. Delayed anaphylaxis to red meat in patients with IgE specific for galactose alpha-1,3-galactose (alpha-gal). Curr Allergy Asthma Rep. 2013;13(1):72-7.
  22. Gonzalez-Quintela A, Dam Laursen AS, Vidal C, Skaaby T, Gude F, Linneberg A. IgE antibodies to alpha-gal in the general adult population: relationship with tick bites, atopy, and cat ownership. Clinical and experimental allergy : journal of the British Society for Allergy and Clinical Immunology. 2014;44(8):1061-8.
  23. Pattanaik D, Lieberman P, Lieberman J, Pongdee T, Keene AT. The changing face of anaphylaxis in adults and adolescents. Annals of allergy, asthma & immunology : official publication of the American College of Allergy, Asthma, & Immunology. 2018;121(5):594-7.
  24. Commins SP, Satinover SM, Hosen J, Mozena J, Borish L, Lewis BD, et al. Delayed anaphylaxis, angioedema, or urticaria after consumption of red meat in patients with IgE antibodies specific for galactose-α-1,3-galactose. Journal of Allergy and Clinical Immunology. 2009;123(2):426-33.e2.
  25. Kennedy JL, Stallings AP, Platts-Mills TAE, Oliveira WM, Workman L, James HR, et al. Galactose- -1,3-galactose and Delayed Anaphylaxis, Angioedema, and Urticaria in Children. PEDIATRICS. 2013;131(5):e1545-e52.
  26. Wilson JM, Schuyler AJ, Workman L, Gupta M, James HR, Posthumus J, et al. Investigation into the α-Gal Syndrome: Characteristics of 261 Children and Adults Reporting Red Meat Allergy. The journal of allergy and clinical immunology In practice. 2019;7(7):2348-58.e4.
  27. Apostolovic D, Rodrigues R, Thomas P, Starkhammar M, Hamsten C, van Hage M. Immunoprofile of α-Gal- and B-antigen-specific responses differentiates red meat-allergic patients from healthy individuals. Allergy. 2018;73(7):1525-31.
  28. Rispens T, Derksen NIL, Commins SP, Platts-Mills TA, Aalberse RC. IgE Production to α-Gal Is Accompanied by Elevated Levels of Specific IgG1 Antibodies and Low Amounts of IgE to Blood Group B. PLoS ONE. 2013;8(2):e55566.
  29. Tripathi A, Commins SP, Heymann PW, Platts-Mills TAE. Delayed Anaphylaxis to Red Meat Masquerading as Idiopathic Anaphylaxis. The Journal of Allergy and Clinical Immunology: In Practice. 2014;2(3):259-65.
  30. Macher BA, Galili U. The Galalpha1,3Galbeta1,4GlcNAc-R (alpha-Gal) epitope: a carbohydrate of unique evolution and clinical relevance. Biochim Biophys Acta. 2008;1780(2):75-88.
  31. Commins SP, James HR, Stevens W, Pochan SL, Land MH, King C, et al. Delayed clinical and ex vivo response to mammalian meat in patients with IgE to galactose-alpha-1,3-galactose. Journal of Allergy and Clinical Immunology. 2014;134(1):108-15.e11.
  32. Rydberg L, Björck S, Hallberg E, Magnusson S, Sumitran S, Samuelsson BE, et al. Extracorporeal ("ex vivo") connection of pig kidneys to humans. II. The anti-pig antibody response. Xenotransplantation. 1996;3(4):340-53.
  33. Ayuso R, Lehrer SB, Tanaka L, Ibanez MD, Pascual C, Burks AW, et al. IgE antibody response to vertebrate meat proteins including tropomyosin. Annals of Allergy, Asthma & Immunology. 1999;83(5):399-405.
  34. Werfel S, Cooke S, Sampson H. Clinical reactivity to beef in children allergic to cow's milk. Journal of Allergy and Clinical Immunology. 1997;99(3):293-300.
  35. Restani P, Ballabio C, Tripodi S, Fiocchi A. Meat allergy. Current Opinion in Allergy and Clinical Immunology. 2009;9(3):265-9.
  36. Crespo JF, Pascual C, Burks AW, Helm RM, Esteban MM. Frequency of food allergy in a pediatric population from Spain. Pediatric allergy and immunology : official publication of the European Society of Pediatric Allergy and Immunology. 1995;6(1):39-43.
  37. Steinke M, Fiocchi A, Kirchlechner V, Ballmer-Weber B, Brockow K, Hischenhuber C, et al. Perceived Food Allergy in Children in 10 European Nations. International Archives of Allergy and Immunology. 2007;143(4):290-5.
  38. Rancé F, Kanny G, Dutau G, Moneret-Vautrin DA. Food hypersensitivity in children: clinical aspects and distribution of allergens. Pediatric allergy and immunology : official publication of the European Society of Pediatric Allergy and Immunology. 1999;10(1):33-8.
  39. Fiocchi A, Restani P, Riva E. Beef allergy in children. Nutrition (Burbank, Los Angeles County, Calif). 2000;16(6):454-7.
  40. Restani P, Fiocchi A, Beretta B, Velonà T, Giovannini M, Galli CL. Meat allergy: III—Proteins involved and cross-reactivity between different animal species. Journal of the American College of Nutrition. 1997;16(4):383-9.
  41. Kanny G, De Hauteclocque C, Moneret-Vautrin D-A. Food anaphylaxis to bovine serum albumin. Journal of Allergy and Clinical Immunology. 1998;101(1):137-9.
  42. Fuentes Aparicio V, Sánchez Marcén I, Pérez Montero A, Baeza ML, de Barrio Fernández M. Allergy to mammal's meat in adult life: immunologic and follow-up study. Journal of investigational allergology & clinical immunology. 2005;15(3):228-31.
  43. Hilger C, Kohnen M, Grigioni F, Lehners C, Hentges F. Allergic cross-reactions between cat and pig serum albumin. Study at the protein and DNA levels. Allergy. 1997;52(2):179-87.
  44. Posthumus J, James HR, Lane CJ, Matos LA, Platts-Mills TAE, Commins SP. Initial description of pork-cat syndrome in the United States. Journal of Allergy and Clinical Immunology. 2013;131(3):923-5.
  45. Fischer J, Hebsaker J, Caponetto P, Platts-Mills TAE, Biedermann T. Galactose-alpha-1,3-galactose sensitization is a prerequisite for pork-kidney allergy and cofactor-related mammalian meat anaphylaxis. Journal of Allergy and Clinical Immunology. 2014;134(3):755-9.e1.
  46. Orhan F, Sekerel BE. Beef allergy: a review of 12 cases. Allergy. 2003;58(2):127-31.
  47. García BE, Lizaso MT. Cross-reactivity syndromes in food allergy. Journal of investigational allergology & clinical immunology. 2011;21(3):162-70; quiz 2 p following 70.