Type:
Whole Allergen
Whole Allergen
Whole Allergen
Halibut
f303
Pleuronectidae
Hippoglossus hippoglossus
Halibut
XM31E6
(ICD-11 is currently under implementation by WHO and the ICD-11 codes displayed in the encyclopedia may not yet be available in all countries)
Skin-sensitivity, anaphylaxis and other symptoms related to food allergy have been reported.
A large, oceanic flat fish sometimes weighing several hundred kilos. The larger fish can be 30 to 35 years old. Halibut is most often frozen but is sold fresh in markets near the fishing areas.
Sera from 2 patients with anaphylactic sensitization to halibut and tuna were studied to assess cross-reactivity with 16 other fish species (1). The IgE binding patterns were more similar as the taxonomical relationship became closer. Skin sensitivity to halibut has been reported (2). Repeated episodes of food-allergic symptoms and anaphylaxis after eating halibut were reported in an early study (3)
Species within groups of fish, like Gadiformes (examples: codfish and hake) and Scombroid fishes (examples: mackerel and tuna) seem to share allergenic components. The overlap of allergen specificity between the groups seems to be moderate or even small.
Last Reviewed- April 2022
Before you proceed ...
We noticed you may be visiting a version of our website that doesn’t match your current location. Would you like to view content based on your region?
Contact us
Interested in utilizing our solutions in your laboratory or healthcare practice?
Not a lab professional or healthcare provider but would like to learn more about allergy testing?
Visit Allergy InsiderChoose your preferred language
Please note: By selecting a different language, you are choosing to view another site. Product availability and indications may vary from what is approved in your region.