Recent searches Clear History
Kontaktieren Sie uns
Allergen Encyclopedia
Table of Contents
Allergengesamtextrakt
f8 Maize/Corn
f8 Maize/Corn Scientific Information
Typ:
Whole Allergen
Anzeigename:
Maize/Corn
Route of Exposure:
Ingestion
Family:
Poaceae (Gramineae)
Species:
Zea mays
Latin Name:
Zea mays
Other Names:
Sweet Corn, Indian Corn, Field Corn
Summary
Maize or corn (Zea mays L.) is a high-yielding and an essential food crop after wheat and rice, available globally. Maize is widely cultivated throughout the world, including countries such as northern Europe, Russia, South Africa, East Asia, the Himalayas, China, Southeast Asia, the Pacific Islands, Chile, and Canada. Allergy to maize has commonly been reported in countries, such as southern Europe and Mexico, where its consumption is popularly high. Oral ingestion of maize can lead to IgE-mediated allergic reactions, like oral food allergy syndrome and even severe reactions, like anaphylaxis. Additionally, inhalation of maize flour at work, can lead to occupational asthma and rhino-conjunctivitis, while skin contact with it, can provoke allergic reactions, such as contact urticaria. Five allergenic components of maize, such as Zea m 1, Zea m 8, Zea m 12, Zea m 14, and Zea m 25 have been identified; among them Zea m 1 and Zea m 14 are the major allergens. Zea m 14 (non-specific lipid transfer protein) is a highly resistant protein, capable of withstanding heat, enzymatic, and chemical treatments. Maize has been found to cross-react with other cereals, such as rice, wheat, barley, etc. Besides this, maize has also shown to cross-react with fruits, such as peach, cherry, cowpea, apricot, grapes as well as pollens, such as grass and Platanus pollens.
Allergen
Nature
Maize or corn (Zea mays L.) is a high-yielding crop and an important source of food, available globally (1). It is considered as an essential food crop, after wheat and rice (2). It is a tall (2-3 m height), monoecious, annual plant, that possesses male and female inflorescences (flower), separately on the same plant. The male inflorescence consists of staminate spikelets that terminates into a panicle, known as tassel, whereas the female pistillate inflorescence arises from the leaf axil as short lateral branch, and is known as the ‘ear’ (2, 3). It bears spikelets in pairs and thus, the ear always has rows of kernels in even numbers (2). The male staminate inflorescence produces the pollens of maize, and the pollination occurs through wind. Usually the pollens are viable within 10 to 30 minutes of shedding; however the viability may rise with favorable environmental conditions (3). Maize is used as a food crop (maize grain) in most of the developing countries, however, it is also used as a fodder, as well as for production of ethanol, in countries, such as USA and Brazil (2, 3).
Harvesting of maize is generally suggested, when the moisture content of the grain is low (15-25%), to allow shelling (4). The name ‘Maize’ was derived from the Spanish word ‘maíz’, that is the name of the plant. It is generally known as Indian corn or corn, in USA, whereas it is known as millies in South Africa (2).
Habitat
Maize is a crop that prefers warm and dry condition. It grows well in the region, where there is moist and frost-free climate (2). This essential crop can be cultivated in a wide range of conditions, ranging from 0-3800 m, above sea level to below 200-2000 mm of precipitation levels. Furthermore, well-aerated, fertile soil helps in producing good quality of crops (1).
Taxonomy
Taxonomic tree of Maize (5)  
Domain Eukaryota
Kingdom Viridiplantae
Phylum Streptophyta
Class Magnoliopsida
Order Poales
Family Poaceae
Genus Zea
Species Zea mays
Tissue
Oral ingestion of maize kernels can cause IgE-mediated allergic reactions (6, 7). The major allergen of maize, a 9 kDa lipid transfer protein (LTP) is a true food allergen, capable of inducing primary sensitization and provoking severe allergic reactions (8).
Epidemiology
Worldwide distribution
Allergy to maize has commonly been reported in countries, such as southern Europe and Mexico, where the consumption of maize is popularly high (9). However, presently maize allergy has been receiving large attention, following its use as a genetically-modified crop in human food (10).
Prevalence of maize allergy was identified in a study that involved 24 Italian and 3 Swiss patients (n=27) with suspected allergy to maize. All the patients (100%) confirmed positive result to skin prick test (SPT) and also showed specific immunoglobulins E (sIgE) for maize (10).
In a study conducted on 661 Mexican patients with a history of allergy, 56 patients (8.7%) reported allergy to maize (11).
Racial and ethnic differences in food allergy phenotype, among children with food allergy in the US have been characterized. A large, retrospective cohort study conducted in US, involved 285 African American (AA), 99 Hispanics, and 433 white children (age range: 0 to 17 years) with a history of food allergy. According to the results, the prevalence of allergy to corn, shellfish, and fish was significantly higher (p<0.01) in AA and Hispanics than in whites. Furthermore, the prevalence of corn allergy in whites, AA, and Hispanics was found to be 2.1%, 15.1%, and 7.1%, respectively (12).
In a survey conducted among school children in France, the prevalence of food allergy was studied using questionnaires. Out of 3500 questionnaires being distributed in 150 classes in 8 schools, 2716 (77.6%) responses were obtained. It was reported that only two out of 244 cases (0.8%) showed allergy to corn, carrots, grapes, and mussels (13).
National Health and Nutrition Examination Survey (NHANES) conducted a periodic survey on the prevalence of different food allergies among 20,686 US children and adolescents. The prevalence of corn allergy obtained in the study was 0.28% among children and 0.22% among adults (14, 15).
An extensive study was conducted on 145 children with atopic dermatitis (AD), who showed positive SPT result to one or more cereal grains, such as corn, wheat, rye, barley, oats, rice. Among 145 patients subjected to food challenges, 21.3% showed positive reactions (31 out of 145 patients), while 25 of these 31 patients (80%) reported positive challenge responses to single grain. Further, it was found that 12% of patients showed a positive response to only corn (3 out of 25 patients) (16).
In a study conducted on 689 allergic patients in Pakistan (Rawalpindi and Islamabad), 16% of patients reported sensitization to corn, based on SPT (110 out of 689 patients) (17).
Risk factors
Respiratory allergic symptoms, such as acute rhinitis and asthma may occur due to occupational exposure to corn flour, in millers, food workers, and homemakers (11). Moreover, the major maize allergen, 9 kDa LTP, shows a high level of sequence homology with the LTP of rice, which attributes to a higher risk of co-sensitization between the individuals with maize allergy and rice-induced allergy (18).
Environmental Characteristics
Worldwide distribution
Maize is extensively cultivated throughout the world, including countries such as northern Europe, Russia, South Africa, East Asia, the Himalayas, China, Southeast Asia, the Pacific Islands, Chile, and Canada. It is considered as a staple food in various tropical regions, especially in Asia, Africa, and Latin America (2).
Though the origin of maize has been controversial, a popular opinion states that it was originated in Mesoamerica, mainly in Mexico and the Caribbean, and later diversified into North and South America, Spain, and then all over the world. The ability of maize to grow in a variety of habitats has helped it to spread quickly and is now grown in more than 100 countries (2).
USA is considered the world’s largest producer of maize, accounting for 42% of the total production. This is followed by countries, such as China (19.4%), Brazil (6.6%), Mexico (3%), and Argentina (2.7%) (2).
Route of Exposure
Main
Oral ingestion of maize can lead to IgE-mediated allergic reactions, like oral food allergy and even severe reactions like anaphylaxis (6, 7).  
Secondary
Inhalation or handling of maize flour at work can lead to occupational asthma and rhino-conjunctivitis. Moreover, skin contact with maize can provoke allergic reactions, such as AR and contact urticaria (6, 7).
Clinical Relevance
Oral allergy symptoms and Anaphylaxis
Food allergy symptoms, such as OAS, urticaria, and anaphylaxis, following consumption of maize, have been reported in some countries, especially in Italy, where polenta (cooked maize flour) is popularly consumed (10).
In a study conducted on 22 patients (age range: 12 to 50 years), 100% of patients suffered from severe systemic reactions, such as anaphylaxis, after consuming maize in different forms (e.g. popcorn, polenta, crisps, etc.). Nine of these 22 patients (41%) presented with OAS (including 5 with localized OAS), before the onset of systemic symptoms, such as laryngeal edema anaphylaxis, and shock. Further, 3 of the 22 patients (13.6%) presented with food-dependent, exercise-induced anaphylaxis (FDEIA) (18).
In a study conducted on 661 Mexican patients with a history of allergy, 56 (8.4%) patients reported maize allergy. Among them, two patients reported to develop urticaria or even angioedema, upon consumption of maize, in forms such as corn-on-the-cob or popcorn. (11).
A double-blind, placebo-controlled food challenge (DBPCFC) was conducted on 16 corn-allergic patients with positive SPT. It was reported that 37.5% of patients (6 out of 16) showed symptoms of itching and urticaria, upon ingestion of cooked corn (19).
In another DBPCFC study involving 27 suspected maize-allergic patients, OAS was found to be the most frequent symptom observed in 53.8% of patients (7 out of 13) with positive findings to maize. Other symptoms included generalized and persistent erythema (23%; 3 out of 13), abdominal pain or diarrhea (23%; 3 out of 13), generalized pruritus (15.3%; 2 out of 13), angioedema (15.3%; 2 out of 13), and one case (7.6%; 1 out of 13) each of generalized urticaria, persistent cough, nausea or vomiting, and eczema flare (10).
A study was conducted on 82 Italian patients (age range: 9 to 47 years) with a history of at least one episode of FDEIA linked with physical exercise (done within 4 hours after a meal), to assess the actual allergenic molecules responsible for FDEIA. Maize and peanut were found to be the most common positive foods (n= 70), followed by apple, peach, onion, lentil and fennel  (20).
Two case studies have been identified that reported allergic reactions, such as OAS and anaphylaxis, after ingestion of corn, in different forms. The first case was of a 44-year-old woman, who had a history of anaphylactic reaction (tingling sensation) post ingestion of corn (licking the cornmeal from fingers). It was further reported that, on subsequent exposure to corn, the women developed urticaria, pruritus, diarrhea, vomiting, and breathlessness. In a two phase, DBPCFC conducted with corn flour, the woman presented with anaphylaxis (21). In the second case, studied on a 34-year-old male, ingestion of baby cereal formula, containing nongluten rice and corn, led to severe reactions, such as sudden respiratory and gastrointestinal (GI) symptoms, reduced consciousness, and paleness. The patient’s history showed that, although he well-tolerated bread and other cereal-derived products, he experienced OAS after taking beer and mustard. Further, it was reported that the patient suffered from occupational asthma, after coming in contact with the rye flour, that was added in the wood boards. The SPT was found to be positive for mustard and several other food allergens. According to DBPCFC conducted with cereal baby food in lime juice, positive reactions, such as pharyngeal itching, cough, and GI symptoms were noted (22).
Allergic rhinitis, rhino-conjunctivitis, and asthma
Respiratory symptoms, such as allergic rhinitis (AR), rhino-conjunctivitis and asthma/occupational asthma have commonly been reported in individuals, who either ingest or come in contact with maize. (6).
In a study conducted on 661 adult patients in Mexico, 8.4% (n=56) of patients presented with a history of maize allergy. Of these 56 individuals, 89.3% (n=50) individuals, who worked with the cereal, experienced AR and/or asthma, after coming in contact with maize. The rest of the  10.7% (n=6) individuals, who reported no history of working with the cereal, presented with asthma and/or AR, upon ingestion of maize (popcorn or corn-on-the- cob) (11).
In a case study of a 40-year-old non-smoker male, occupational exposure to maize while working on a maize extrusion process led to rhino-conjunctivitis and asthma. These allergic reactions lasted for around 6 months while at work, however the symptoms reduced, when off-work. The SPT results was found to be positive for maize flour, wheat flour, and beer. Moreover, the specific IgE determinations showed positive result for maize flour extracts, however it was reported negative for other cereal flours (6).
Two more case studies documented about corn-induced occupational asthma (6). The first case was of a 24-year-old, non-smoker male who reported a history of rhinorrhea, sneezing, and chest tightness, as a result of handling corn dust for making stock feed. The male informed that he did not experience these symptoms in the past, however, his symptoms got worsened on handling the corn dust. The bronchoprovocation test and Enzyme Linked Immunosorbent Assay (ELISA) suggested that corn dust can lead to IgE-mediated bronchoconstriction in an exposed individual (23). In the second case,a 19-year-old girl, with no prior history of seasonal or perennial allergies, developed erythematous itching rash, on both hands as well as face, along with dyspnea. She worked in a pharmaceutical company that manufactured tablets. Every day, after about 2 hours of working on tablets (containing maize starch, microcrystalline cellulose and lactose), she developed allergic symptoms. A strong positive reaction (including sneezing, conjunctival reaction, and pruritus in nose and eye) was elicited within 6 minutes of nasal provocation test with maize bran. The study revealed that the patient developed allergy to maize through inhalation and cutaneous contact to the allergen, while at work. (9).
Maize has also been reported as a causative agent in baker’s asthma. These patients, however, have never reported problems after consumption of this cereal grain (10)
Atopic dermatitis
A 33-year-old nurse with a history of asthma and seasonal AR, presented with hand dermatitis, since the time she started nursing. She complained of having intermittent, itchy blisters on her hands, which improved when she was off work. She also experienced ‘itchy throat’, when she ate bananas, kiwi, avocado, and cantaloupe. The SPT showed positive response for corn, and the nurse was advised to avoid wearing gloves that contain corn-starch powder. It was reported that her symptoms started improving, after she used a non-cornstarch-based gloves (24).
Prevention and Therapy
Prevention strategies
Avoidance
Corn allergy can be managed by strictly avoiding the corn ingredients. Moreover, corn is considered as one of the most essential, genetically modified (GM) crops, and hence an appropriate identification of each GM ingredient is needed for GM food labeling as well as monitoring. Furthermore, proper information about the presence of corn in processed foods should be given, so as to make people aware of quality, safety, and food accuracy (25).
Molecular Aspects
Allergenic molecules
The World Health Organization (WHO) and International Union of Immunological Societies’ (IUIS) Allergen Nomenclature Subcommittee have registered five allergenic components, i.e., Zea m 1, Zea m 8, Zea m 12, Zea m 14 and Zea m 25 for maize/corn (26). Among the five maize allergenic molecules, Zea m 8 and Zea m 14 are exposed through ingestion, whereas Zea m 1, Zea m 12, and Zea m 25 are exposed through inhalation (26).
Allergen Biochemical Name  Molecular Weight (kDa) Allergenicity
Zea m 1 β-expansin 25-35
  • Major maize pollen allergen (8)
  • All 12 sera from grass pollen allergic patients revealed IgE binding to Zea m 1 on immunoblot assay (26)
Zea m 8 Class IV chitinase
28.6
  • Maize endochitinase showed 62% sequence homology with grape endochitinase IV (7)
  • This sequence homology leads to cross-reactivity between maize and  grape endochitinase (7)
Zea m 12 Profilin
14
  • Panallergen profilin (8)
  • Shows cross-reactivity with wheat profilin, when tested (ELISA and Western blot) from serum pool of patients with wheat and maize allergy (26).
  • Heat-sensitive, and hence unable to produce sensitization through GI tract (8).
Zea m 14 Nonspecific lipid-transfer protein 1 9
  • Major allergen. Highly heat-stable protein allergen of maize (18)
  • 19 out of 22 sera (86%)  of maize-allergic patients found Zea m 14 allergen on immunoblot assay (26).
Zea m 25 Thioredoxin
13.8
  • Minor allergen (7).
  • Regulatory protein that lowers intrachain disulphide bridges of target proteins (8).
  • High cross-reactivity found with Tri a 25 (wheat thioredoxin), among bakers having occupational asthma (74%) (26).
kDa: kilodaltons, IgE: Immunoglobulin E, ELISA: Enzyme-linked immunosorbent assay
Besides the main allergens listed above, there are some other allergens found in maize, that cause allergic reactions. Zea m 20S, belonging to pectate lyase family has been reported as an allergen present in maize seeds. Two other maize allergens, that belong to prolamin superfamily, i.e., Zea m 27 kDa Zein and Zea m 50 kDa Zein, have also been found in maize seeds. Further, a vicilin-like embryo storage protein, known as Zea m G1, have reported to cause allergic reaction on ingestion of seeds (8). In a study, two h-type maize thioredoxins, namely ZmTRXh1 (Zea m 25) and ZmTRXh2, in maize had been identified (27).
A 50 kDa IgE-binding protein band belonging to reduced soluble protein (RSP) fraction was detected by the serum IgE in patients diagnosed with an allergy to corn. This  50 kDa salt-unextractable band was considered to be a candidate allergen in IgE-mediated food allergy to corn products, due to its high resistance towards cooking and enzymatic digestion (19).
The amylase-trypsin inhibitor (16 kDa) is a minor allergen found in maize seed. This protein shows homology with other cereals, belonging to the Poaceae family, and is anticipated to cause baker’s asthma (18).
Biomarkers of severity
The major allergen of maize (Zea m 14) is a highly-resistant protein, capable of withstanding heat, enzymatic, and chemical treatment (7). The protein, if heated to a temperature as high as 100oC for 160 minutes, remains unaffected. Moreover, owing to its ability to resist proteolytic digestion and food processing, it can reach the GI immune system, resulting in sensitization and triggering of systemic symptoms (8). Furthermore, it is an extremely concentrated protein that can be isolated quickly; hence can be considered as an ideal candidate for a food allergen (7). 
Cross-reactivity
Maize has been found to cross-react with other cereals, such as rice, wheat, barley, etc. Lipid transfer protein is considered as a major allergen of maize, that has shown sequence identity with LTP of rice, wheat, and barley (18). A study revealed that IgE antibodies from 28.5% of patients with maize allergy (n=7) showed cross-reactivity with a 56 kDa rice allergen (28). In another study, rice extracts completely inhibited all the maize allergens. Moreover, maize LTP has been found to have 79% identity and 92% similarity with rice LTP. The identity was found lower in case of wheat (59%-79%) and barley (57%-78%). Furthermore, the inhibitory potential of wheat, barley, and grass pollen extracts showed complete inhibition to 16 kDa protein and other higher molecular weight protein bands, from the sera of patients allergic to maize. It was demonstrated that these three cereals showed cross-reactivity with trypsin/activated Hageman factor (XIIA) inhibitor and other proteins of maize (18).
Additionally, maize LTP has shown sequence similarity with certain fruits. In a case report of a 19-year-old boy, peach LTP was found to be cross-reactive to corn, along with rice, apple, walnut, hazelnut, and peanut, with the frequency of cross reactivity being 94%, 93%, 97%, 20%, 66%, and 91% respectively (29). A study found LTP from maize showing 63% sequence similarity with LTP of apricot (Mr 9000) (30). Another study reported high level of amino acid sequence identity (59.3%) between LTP from cherry and LTP from maize. Cowpea also showed high sequence homology with the LTP from maize (72%) (31).
Besides LTP, another maize allergen also showed sequence homology with cereal allergen, such as wheat allergen. In a study that assessed sequence identity between two maize thioredoxins (Zea m 25 and ZmTRXh2) and wheat thioredoxin allergen (Tri a 25), the analysis showed 74% of sequence identity between Zea m 25 and Tri a 25 allergen. Furthermore, low sequence identity was reported between ZmTRXh2 and Tri a 25 (46%) (27).
The pollen of Platanus aceriflora (P. aceriflora) showed cross-reactivity with certain plant derived foods. Enzyme allergosorbent (EAST) inhibition assays showed 90% inhibition when extracts of corn apple, chickpea, lettuce, peanut or hazelnut were used in solid phase and the inhibitor phase contained P. aceriflora pollen extract (32).
Furthermore, Zea m 8 (allergenic chitinase from maize) and grape (Class IV chitinase), show about 40-50% of sequence similarity with the class I chitinase, present in foods of plant origin (papaya, avocado, chestnut, kiwi, banana etc.) (33). This may account for high rate of cross-reactivity between maize and grape chitinase allergens, resulting in clinical reactions in patients sensitive to maize and grape (7). According to a study conducted in almond-allergic patients, high level of co-sensitization or cross-reactivity was reported between 50 kDa γ-zein (maize) and almond major protein (AMP), whereas, low cross-reactivity was found between 27 kDa γ-zein (maize) and AMP (34).
Compiled By
Author: Turacoz Healthcare Solutions
Reviewer: Dr. Fabio Iachetti
Last reviewed: January 2021
References
  1. Ramirez-Cabral NYZ, Kumar L, Shabani F. Global alterations in areas of suitability for maize production from climate change and using a mechanistic species distribution model (CLIMEX). Sci Rep. 2017;7(1):5910.
  2. Verheye W. Growth and production of maize: traditional low-input cultivation. Encyclopedia of Life Support Systems (EOLSS). Oxford, UK: UNESCO-EOLSS Publishers; 2010.
  3. CFIA. The Biology of Zea mays (L.) (Maize). Ottawa, Ontario: Plant Biosafety Office; 1994.
  4. Ngoune Tandzi L, Mutengwa CS. Estimation of maize (Zea mays L.) yield per harvest area: appropriate methods. Agronomy. 2020;10(1):29.
  5. NCBI. Zea mays  [18-12-2020]. Available from: https://www.ncbi.nlm.nih.gov/Taxonomy/Browser/wwwtax.cgi?mode=info&id=4577.
  6. Guillen D, Barranco P, Palacin A, Quirce S. Occupational Rhinoconjunctivitis due to Maize in a Snack Processor: A Cross-Reactivity Study Between Lipid Transfer Proteins From Different Cereals and Peach. Allergy Asthma Immunol Res. 2014;6(5):470-3.
  7. Pastorello EA, Farioli L, Pravettoni V, Scibilia J, Conti A, Fortunato D, et al. Maize food allergy: lipid-transfer proteins, endochitinases, and alpha-zein precursor are relevant maize allergens in double-blind placebo-controlled maize-challenge-positive patients. Anal Bioanal Chem. 2009;395(1):93-102.
  8. Jose C. Jimenez-Lopez SOK, Emma Gachomo,Antonio Jesús Castro,María Isabel Rodríguez-García,Juande Alche. Molecular Features of Maize Allergens and their Implications in Human Health.  MAIZE: CULTIVATION, USES AND HEALTH BENEFITS. Agriculture Issues and Policies ed: NOVA Publishers; 2012.
  9. Maniu CM, Faupel U, Siebenhaar G, Hunzelmann N. Maize: a new occupational allergen in the pharmaceutical industry. Allergy. 2010;65(7):930-1.
  10. Scibilia J, Pastorello EA, Zisa G, Ottolenghi A, Ballmer-Weber B, Pravettoni V, et al. Maize food allergy: a double-blind placebo-controlled study. Clin Exp Allergy. 2008;38(12):1943-9.
  11. Valencia Zavala MP, Vega Robledo GB, Sanchez Olivas MA, Duarte Diaz RJ, Oviedo CL. Maize (Zea mays): allergen or toleragen? Participation of the cereal in allergic disease and positivity incidence in cutaneous tests. Rev Alerg Mex. 2006;53(6):207-11.
  12. Mahdavinia M, Fox SR, Smith BM, James C, Palmisano EL, Mohammed A, et al. Racial Differences in Food Allergy Phenotype and Health Care Utilization among US Children. J Allergy Clin Immunol Pract. 2017;5(2):352-7 e1.
  13. Rance F, Grandmottet X, Grandjean H. Prevalence and main characteristics of schoolchildren diagnosed with food allergies in France. Clin Exp Allergy. 2005;35(2):167-72.
  14. Messina M, Venter C. Recent Surveys on Food Allergy Prevalence. Nutrition Today. 2020;55(1):22-9.
  15. McGowan EC, Keet CA. Prevalence of self-reported food allergy in the National Health and Nutrition Examination Survey (NHANES) 2007-2010. J Allergy Clin Immunol. 2013;132(5):1216-9 e5.
  16. Jones SM, Magnolfi CF, Cooke SK, Sampson HA. Immunologic cross-reactivity among cereal grains and grasses in children with food hypersensitivity. J Allergy Clin Immunol. 1995;96(3):341-51.
  17. Inam M, Shafique RH, Roohi N, Irfan M, Abbas S, Ismail M. Prevalence of sensitization to food allergens and challenge proven food allergy in patients visiting allergy centers in Rawalpindi and Islamabad, Pakistan. Springerplus. 2016;5(1):1330.
  18. Pastorello EA, Farioli L, Pravettoni V, Ispano M, Scibola E, Trambaioli C, et al. The maize major allergen, which is responsible for food-induced allergic reactions, is a lipid transfer protein. J Allergy Clin Immunol. 2000;106(4):744-51.
  19. Pasini G, Simonato B, Curioni A, Vincenzi S, Cristaudo A, Santucci B, et al. IgE-mediated allergy to corn: a 50 kDa protein, belonging to the Reduced Soluble Proteins, is a major allergen. Allergy. 2002;57(2):98-106.
  20. Romano A, Scala E, Rumi G, Gaeta F, Caruso C, Alonzi C, et al. Lipid transfer proteins: the most frequent sensitizer in Italian subjects with food-dependent exercise-induced anaphylaxis. Clin Exp Allergy. 2012;42(11):1643-53.
  21. Tanaka LG, El-Dahr JM, Lehrer SB. Double-blind, placebo-controlled corn challenge resulting in anaphylaxis. J Allergy Clin Immunol. 2001;107(4):744.
  22. Asensio T, Armentia A, Lombardero M, Callejo A, Martin G, Castrodeza J. Cereal-induced anaphylaxis in an adult after eating a baby cereal formula. Allergol Immunopathol (Madr). 2004;32(5):310-1.
  23. Park HS, Nahm DH. Identification of IgE-binding components in occupational asthma caused by corn dust. Ann Allergy Asthma Immunol. 1997;79(1):75-9.
  24. Liu W, Nixon RL. Corn contact urticaria in a nurse. Australas J Dermatol. 2007;48(2):130-1.
  25. Gabriadze I, Kutateladze T, Vishnepolsky B, Karseladze M, Datukishvili N. Application of PCR-based methods for rapid detection of corn ingredients in processed foods. International Journal of Nutrition and Food Sciences. 2014;3(3):199-202.
  26. WHO/IUIS. Zea mays (Maize) 2019 [18-12-2020]. Available from: http://www.allergen.org/search.php?Species=Zea%20mays.
  27. Weichel M, Vergoossen NJ, Bonomi S, Scibilia J, Ortolani C, Ballmer-Weber BK, et al. Screening the allergenic repertoires of wheat and maize with sera from double-blind, placebo-controlled food challenge positive patients. Allergy. 2006;61(1):128-35.
  28. Krishnan HB, Chen MH. Identification of an abundant 56 kDa protein implicated in food allergy as granule-bound starch synthase. J Agric Food Chem. 2013;61(22):5404-9.
  29. Asero R, Mistrello G, Roncarolo D, de Vries SC, Gautier MF, Ciurana CL, et al. Lipid transfer protein: a pan-allergen in plant-derived foods that is highly resistant to pepsin digestion. Int Arch Allergy Immunol. 2001;124(1-3):67-9.
  30. Conti A, Fortunato D, Ortolani C, Giuffrida MG, Pravettoni V, Napolitano L, et al. Determination of the primary structure of two lipid transfer proteins from apricot (Prunus armeniaca). J Chromatogr B Biomed Sci Appl. 2001;756(1-2):123-9.
  31. Carvalho AO, Souza-Filho GA, Ferreira BS, Branco AT, Araujo IS, Fernandes KV, et al. Cloning and characterization of a cowpea seed lipid transfer protein cDNA: expression analysis during seed development and under fungal and cold stresses in seedlings' tissues. Plant Physiol Biochem. 2006;44(11-12):732-42.
  32. Enrique E, Cistero-Bahima A, Bartolome B, Alonso R, San Miguel-Moncin MM, Bartra J, et al. Platanus acerifolia pollinosis and food allergy. Allergy. 2002;57(4):351-6.
  33. Leoni C, Volpicella M, Dileo M, Gattulli BAR, Ceci LR. Chitinases as Food Allergens. Molecules. 2019;24(11).
  34. Lee SH, Benmoussa M, Sathe SK, Roux KH, Teuber SS, Hamaker BR. A 50 kDa maize gamma-zein has marked cross-reactivity with the almond major protein. J Agric Food Chem. 2005;53(20):7965-70.
  35. Asero R, Mistrello G, Roncarolo D, Amato S, van Ree R. A case of allergy to beer showing cross-reactivity between lipid transfer proteins. Ann Allergy Asthma Immunol. 2001;87(1):65-7.