Recent searches Clear History
Fale conosco
Allergen Encyclopedia
Table of Contents
Alérgeno completo
f10 Sesame seed
f10 Sesame seed Scientific Information
Tipo:
Whole Allergen
Nome para exibição:
Sesame seed
Route of Exposure:
Ingestion
Family:
Pedaliaceae
Species:
Sesamum indicum
Latin Name:
Sesamum indicum
Other Names:
Sesame, Benne seed
Summary
Sesamum indicum is one of the oldest oilseed plants belonging to the family Pedaliaceae and widely grown species in Sesamum genus. Asia and Africa hold 96% of world sesame seed production. It is commonly used in Middle Eastern cuisine and in Mediterranean diet and is increasingly being used in various foods in US like breads, sauces like tahini or confectionaries like halva. The prevalence of sesame allergy in USA, Mexico, Canada and Europe is reported 0.1% and 0.8% in Australia. It is considered ninth most common childhood food allergy in US while third most common food allergen and second most common food to cause anaphylaxis in children in Israel. Australia, Canada, Europe and New Zealand have recognized sesame as priority food allergen which required food labeling, but it is not currently mandatory in USA. Oral ingestion of sesame seeds either whole or in food preparations can lead to symptoms ranging from oral food allergy presented by recurrent episodes of wheezing and dyspnea to asthma, atopic dermatitis to anaphylaxis. Sesame skin prick test was found to be more accurate predictor than sesame serum specific IgE levels. The World Health Organization(WHO)/International Union of Immunological Societies(IUIS) Allergen Nomenclature Subcommittee has registered seven sesame allergen components: four storage proteins including two 2S albumins (Ses i 1 and Ses i 2) and two 11S globulins (Ses i 6 and Ses i 7), sesame extract also contains two oleosins (Ses i 4 and Ses i 5) and one vicilin-like 7S globulin (Ses i 3). Sesame allergy was reported as commonly co-existent in patients with peanut and tree nut (pine nut, macadamia nut, brazil nut and hazelnut) allergies.
Allergen
Nature
Sesamum indicum (S. indicum) is one of the oldest oilseed plants. It is a self-pollinated annual plant grown for its seed which contains 50-60% oil (1, 2). 
Taxonomy
S. indicum is a widely grown species in Sesamum genus. 23 species have been listed under the Sesamum and S. indicum is assumed to be domesticated from S. malabaricum which was native to South Asia and spread to the west through Mesopotamia. S. radiatum is also cultivated in certain African countries as a leafy vegetable (1). 
Taxonomic tree of Sesame (3)  
Domain Eukaryota
Kingdom Plantae
Phylum Spermatophyta
Subphylum Angiospermae
Class Dicotyledonae 
Order Scrophulariales
Family Pedaliaceae
Genus Sesamum
Species Sesamum indicum
Tissue
The proteins present in the water-soluble fraction and oil body fraction of sesame seed extract are identified as allergens (4). Whole sesame seeds and seed oil are said to be better tolerated compared to sesame paste such as tahini or hummus since they contain fewer allergens or are uncrushed and pass through the gut undigested (5). 
Epidemiology
Worldwide distribution
The prevalence of sesame allergy in the USA, Mexico, Canada and Europe is reported 0.1% (6, 7) and 0.8% in Australia (8). Recently, it is considered as the ninth most common childhood food allergy in the US (5) with 0.23% prevalence in a population survey (8). Sesame was responsible for allergy in 9.8% of food allergic patients in the UK as per Pronuts study (9).
It is the third most common food allergen and the second most common food to cause anaphylaxis in children in Israel (10) with a prevalence of 0.93% (11).
Allergy to sesame occurs in individuals ranging from infants to adults including de novo sensitization in adults (12). The age-specific prevalence reported convincing sesame allergy in 0.21% children and 0.24% adults in the US while probable sesame allergy in Canada was found in 0.23% children and 0.05% adults (8).
Risk factors
Sesame allergy has been reported to co-exist in 50-54% of patients with both peanut and tree nut allergy (13). Thus, peanut and tree nut allergies could pose a risk for sesame allergy. 
Environmental Characteristics
Living environment
Sesame adapts well to environmental variations like water and light intensity and grows throughout the year (14). Tropical and temperate climate conditions with stored soil moisture and minimal irrigation or rainfall may produce high-quality yields for sesame seeds under high temperatures (15).
Worldwide distribution
The sesame plant has been found to have its origin in Africa and India (2, 15). Asia and Africa hold 96% of world sesame seed production where it is an important part of the human diet. The country with the highest sesame seed consumption is Tanzania followed by China, Sudan, Myanmar, India, Ethiopia, and Nigeria (16). The black and white sesame varieties are most consumed in Western countries (17). It is commonly used in Middle Eastern cuisine and included in the Mediterranean diet but now increased usage in the US in various foods like bread, sauces like tahini, or confectionaries like halva. Sesame oil is widely used in the pharmaceutical and cosmetic industry in addition to the food industry (12). 
Route of Exposure
Main
Oral ingestion of sesame seeds either whole or in food preparations can lead to symptoms ranging from oral food allergy to atopic dermatitis to anaphylaxis (12, 18, 19).
Secondary
Certain cases have reported allergic symptoms like contact dermatitis due to mucosal contact or severe respiratory symptoms due to inhalation (20).
Detection
Main methods
Koppelman, Söylemez (17) developed a sensitive ELISA for detection and quantification of sesame seed residue in food with a lower limit of 0.5 ppm and no cross-reactivity with 92 other food or food ingredients. This may be helpful for detection in highly allergic individuals.
Clinical Relevance
Sesame allergy can manifest symptoms ranging from oral food allergy syndrome presented by recurrent episodes of wheezing and dyspnea to atopic dermatitis and anaphylaxis (12).
Anaphylaxis and food allergy
Allergic manifestations in patients with sesame allergy found skin symptoms to be most common followed by gastrointestinal, respiratory and cardiovascular symptoms along with anaphylaxis in few patients (18, 19). However, among several nuts evaluated, sesame was found to produce allergic symptoms with the highest severity and more involvement of the lower respiratory tract or cardiovascular/neurological system (9). 2.8% of cases in Canada and 43% in Israel reported sesame induced anaphylaxis among the total cases of food-induced anaphylaxis (21). Anaphylactic symptoms were prominent in sesame allergic patients as seen from studies reporting 41% in Japan (19) and 61.7% in North America and Canada (22). Further, 5 cases of food protein-induced enterocolitis syndrome (non-IgE mediated hypersensitivity) induced by sesame in children in Israel were reported, presented as acute repetitive vomiting, lethargy, and pallor (21).
Allergic rhinitis
A study conducted in the UK on 33 adult patients (16-81 years) who underwent sesame oral food challenge (OFC) reported allergic rhinitis in 48% of patients (23).
Asthma
Asthma was reported in 28.3% of sesame allergic children in Japan (19) while in 36% of UK adults who underwent OFC (23). It was also reported in 25.4% of patients with positive sesame OFC in Israel (18). Bronchospasm was reported in 2 cases of bakers and a young girl due to sesame seeds through inhalation route (20).
Atopic dermatitis
Li, Gunawardana (23) reported atopic dermatitis in 30% of patients in the UK study while Maruyama, Nakagawa (19) reported in 66.3% sesame allergic Japanese children. Atopic dermatitis was found to be positively associated with positive sesame OFC with 56.7% of patients affected having positive sesame OFC (18). Lignin like allergens, sesamol and sesamin in sesame oil has been identified to cause allergic contact dermatitis (20).
Other diseases
Contact urticaria was reported in one case to sesame by oral mucosal contact manifested as oral edema, erythema, and blood-tinged vesicles. Delayed hypersensitivity as contact dermatitis was also reported in a patient who received progesterone injection in sesame oil manifested as respiratory symptoms several days after receiving the injection (20).
Diagnostics Sensitization
Skin prick tests
 Sesame skin prick test (SPT) was found to be a more accurate predictor than sesame serum specific IgE (24) while Ses i 1 IgE levels were found better than SPT in another study (5, 25).
Prevention and Therapy
Other topics
Australia, Canada, Europe and New Zealand have recognized sesame as a priority food allergen that required food labeling, but it is not currently mandatory in the USA (8). Sesame food allergy has been shown to resolve in 20-30% of patients (5).
Molecular Aspects
Allergenic molecules
The World Health Organization(WHO)/International Union of Immunological Societies(IUIS) Allergen Nomenclature Subcommittee has registered seven sesame allergen components: four storage proteins including two 2S albumins (Ses i 1 and Ses i 2) and two 11S globulins (Ses i 6 and Ses i 7), sesame extract also contains two oleosins (Ses i 4 and Ses i 5) and one vicilin-like 7S globulin (Ses i 3).
Allergen
Biochemical Name
Molecular weight
Allergenicity
Ses i 1
2S albumin
9 kDa
All sesame allergic patients were found sensitized to 2S albumins  and with positive IgE binding (19, 26). Ses i 1 and Ses i 2 sensitization was found in 91.7% and 66.7% sesame allergic symptomatic Japanese patients with Ses i 1 to be more allergenic (19).
Related to peanut allergen Ara h 2,6 and 7 (27).
Ses i 2
2S albumin
7 kDa
Ses i 3
7S vicilin-like globulin
45 kDa
75% sesame allergic patients revealed obvious IgE binding (19)
Related to peanut allergen Ara h 1(27).
Ses i 4
Oleosin
17 kDa
Consistent sensitization was demonstrated in 29 of 32 sesame allergic patients showing systemic reactions (28).
Related to peanut allergens Ara h 12, 13, 14 and 15 (27).
Ses i 5
Oleosin
15 kDa
Ses i 6
11S globulin
52 kDa
The basic subunits of these proteins were found to be major allergens in a cohort of 18 Italians (3-73 years) (29). 67% sesame allergic patients showed IgE binding (19).
Related to peanut allergens Ara h 3 (27).
Ses i 7
11S globulin
57 kDa
Biomarkers of severity
Ehlers, Rossnagel (31) identified sesame oleosins as minor allergens but suggested their ineffectiveness in diagnosing sesame allergy based on serum IgE and IgG detection. However, oleosin is identified as dominant allergen in 5 of the 6 patients who reported anaphylaxis to sesame (10). 
Cross-reactivity
Sesame allergy was reported as commonly co-existent in patients with peanut and tree nut (pine nut, macadamia nut, brazil nut and hazelnut) allergies (9, 13, 32). Cross reactivity and co-sensitizations have been reported among allergens in sesame and other foods like peanuts, hazelnut, walnut, black walnut, cashew, macadamia, pistachio, kiwi, rye and poppy seeds (5, 12, 20).
IgE cross-reactivity between 7S and 11S globulins from sesame and peanuts is common. The homology between peanut 7S globulin (Ara h 1) and 11 S globulin (Ara h 3) and sesame proteins is high with Ses i 3 showing 36% homology to Ara h 1while IgE binding epitopes of peanut 2S albumin (Ara h 2) has limited sequence identity with sesame 2S albumin (Ses i 1) (19). This may give false positive sesame IgE based diagnosis (13). Further, Albunni, Wessels (33) reported cross-sensitization between chia seeds and sesame seeds by demonstrating the strong inhibition of anti-sesame antibodies by chia globulin fraction. 
Compiled By
Author: Turacoz Healthcare Solutions
Reviewer: Dr. Magnus Borres
Last reviewed: November 2020
References
  1. Dossa K, Diouf D, Wang L, Wei X, Zhang Y, Niang M, et al. The Emerging Oilseed Crop Sesamum indicum Enters the "Omics" Era. Front Plant Sci. 2017;8:1154.
  2. Dossa K, Wei X, Zhang Y, Fonceka D, Yang W, Diouf D, et al. Analysis of Genetic Diversity and Population Structure of Sesame Accessions from Africa and Asia as Major Centers of Its Cultivation. Genes (Basel). 2016;7(4).
  3. CABI. Sesamum indicium (Sesame) Wallingford, UK: CAB International; 2019 [updated Octber 30, 2020].
  4. Leduc V, Moneret-Vautrin DA, Tzen JTC, Morisset M, Guerin L, Kanny G. Identification of oleosins as major allergens in sesame seed allergic patients. Allergy. 2006;61(3):349-56.
  5. Sokol K, Rasooly M, Dempsey C, Lassiter S, Gu W, Lumbard K, et al. Prevalence and diagnosis of sesame allergy in children with IgE-mediated food allergy. Pediatr Allergy Immunol. 2020;31(2):214-8.
  6. Bedolla-Barajas M, Bedolla-Pulido TR, Macriz-Romero N, Morales-Romero J, Robles-Figueroa M. Prevalence of Peanut, Tree Nut, Sesame, and Seafood Allergy in Mexican Adults. Rev Invest Clin. 2015;67(6):379-86.
  7. Sicherer SH, Muñoz-Furlong A, Godbold JH, Sampson HA. US prevalence of self-reported peanut, tree nut, and sesame allergy: 11-year follow-up. The Journal of allergy and clinical immunology. 2010;125(6):1322-6.
  8. Warren CM, Chadha AS, Sicherer SH, Jiang J, Gupta RS. Prevalence and Severity of Sesame Allergy in the United States. JAMA Netw Open. 2019;2(8):e199144.
  9. Brough HA, Caubet JC, Mazon A, Haddad D, Bergmann MM, Wassenberg J, et al. Defining challenge-proven coexistent nut and sesame seed allergy: A prospective multicenter European study. J Allergy Clin Immunol. 2020;145(4):1231-9.
  10. Adatia A, Clarke AE, Yanishevsky Y, Ben-Shoshan M. Sesame allergy: current perspectives. J Asthma Allergy. 2017;10:141-51.
  11. Garkaby J, Epov L, Musallam N, Almog M, Bamberger E, Mandelberg A, et al. The Sesame-Peanut Conundrum in Israel: Reevaluation of Food Allergy Prevalence in Young Children. J Allergy Clin Immunol Pract. 2020.
  12. Dalal I, Goldberg M, Katz Y. Sesame seed food allergy. Curr Allergy Asthma Rep. 2012;12(4):339-45.
  13. Tuano KT, Dillard KH, Guffey D, Davis CM. Development of sesame tolerance and cosensitization of sesame allergy with peanut and tree nut allergy in children. Ann Allergy Asthma Immunol. 2016;117(6):708-10.
  14. Zehra S, Koaser R, Ahmad Z, Naz N, Atta M, Satti S, et al. Screening of different varieties of sesame (Sesamum indicumL) for growth,yield and oil response under warm climatic conditions. 2017.
  15. Ali Al-Somain BH, Migdadi HM, Al-Faifi SA, Alghamdi SS, Muharram AA, Mohammed NA, et al. Assessment of genetic diversity of sesame accessions collected from different ecological regions using sequence-related amplified polymorphism markers. 3 Biotech. 2017;7(1):82.
  16. Myint D, Gilani S, M. K, KN. W. Sustainable Sesame (Sesamum indicum L.) Production through Improved Technology: An Overview of Production, Challenges, and Opportunities in Myanmar. Sustainability. 2020;12(9):3515.
  17. Koppelman SJ, Söylemez G, Niemann L, Gaskin FE, Baumert JL, Taylor SL. Sandwich Enzyme-Linked Immunosorbent Assay for Detecting Sesame Seed in Foods. Biomed Res Int. 2015;2015:853836.
  18. Epov L, Garkaby J, Almog M, Ben-Or O, Schichter-Konfino V, Toker O, et al. Using skin prick test to sesame paste in the diagnosis of sesame seed allergy. J Allergy Clin Immunol Pract. 2020;8(4):1456-8.
  19. Maruyama N, Nakagawa T, Ito K, Cabanos C, Borres MP, Movérare R, et al. Measurement of specific IgE antibodies to Ses i 1 improves the diagnosis of sesame allergy. Clin Exp Allergy. 2016;46(1):163-71.
  20. Patel A, Bahna SL. Hypersensitivities to sesame and other common edible seeds. Allergy. 2016;71(10):1405-13.
  21. Ovadia A, Nahum A, Tasher D, Abiri S, Epov L, Kessel A, et al. Sesame: An unrecognized trigger of food protein-induced enterocolitis syndrome. J Allergy Clin Immunol Pract. 2019;7(1):305-6.
  22. Segal L, Ben-Shoshan M, Alizadehfar R, Primeau MN, Asai Y, Killorn KR, et al. Initial and accidental reactions are managed inadequately in children with sesame allergy. J Allergy Clin Immunol Pract. 2017;5(2):482-5.
  23. Li PH, Gunawardana N, Thomas I, Ue KL, Siew L, Watts TJ, et al. Sesame allergy in adults: Investigation and outcomes of oral food challenges. Ann Allergy Asthma Immunol. 2017;119(3):285-7.
  24. Saf S, Sifers TM, Baker MG, Warren CM, Knight C, Bakhl K, et al. Diagnosis of Sesame Allergy: Analysis of Current Practice and Exploration of Sesame Component Ses i 1. J Allergy Clin Immunol Pract. 2020;8(5):1681-8.e3.
  25. Salari F, Bemanian MH, Fallahpour M, Tavakol M, Shokri S, Baniadam L, et al. Comparison of Diagnostic Tests with Oral Food Challenge in a Clinical Trial for Adult Patients with Sesame Anaphylaxis. Iran J Allergy Asthma Immunol. 2020;19(1):27-34.
  26. Asero R, Cecchi L, Cervone M, Crivellaro M, Lodi Rizzini F, Pravettoni V, et al. Detection of 20 kDa and 32 kDa IgE-binding proteins as the major allergens in Italian sesame seed allergic patients. Eur Ann Allergy Clin Immunol. 2014;46(1):22-5.
  27. Ziegler JB, Aalberse RC. Sesame: An Increasingly Popular Word and Common Food Allergen. J Allergy Clin Immunol Pract. 2020;8(5):1689-91.
  28. Jappe U, Schwager C. Relevance of Lipophilic Allergens in Food Allergy Diagnosis. Curr Allergy Asthma Rep. 2017;17(9):61.
  29. Magni C, Ballabio C, Restani P, Fuggetta D, Alessandri C, Mari A, et al. Molecular insight into IgE-mediated reactions to sesame (Sesamum indicum L.) seed proteins. Ann Allergy Asthma Immunol. 2010;105(6):458-64.
  30. Jung TD, Choi SI, Choi SH, Cho BY, Sim WS, Lee SJ, et al. Changes in the Anti-Allergic Activities of Sesame by Bioconversion. Nutrients. 2018;10(2).
  31. Ehlers AM, Rossnagel M, Brix B, Blankestijn MA, Le TM, Suer W, et al. Sesame oleosins are minor allergens. Clin Transl Allergy. 2019;9:32.
  32. Anagnostou A. Insights into tree nut and sesame consumption from a cohort of 80 peanut-allergic children. Pediatr Allergy Immunol. 2019;30(3):389-92.
  33. Albunni BA, Wessels H, Paschke-Kratzin A, Fischer M. Antibody Cross-Reactivity between Proteins of Chia Seed ( Salvia hispanica L.) and Other Food Allergens. J Agric Food Chem. 2019;67(26):7475-84.