Recent searches Clear History
Fale conosco
Allergen Encyclopedia
Table of Contents
Componente
f458 Ara h 18
f458 Ara h 18 Scientific Information
Tipo:
Component
Name; WHO/IUIS:
Ara h 18
Route of Exposure:
Ingestion
Biological function:
Cyclophilin - peptidyl-prolyl cis-trans isomerase
Allergen code:
f458
Source Material:
Peanut extract
Other Names :
Cyclophilin - peptidyl-prolyl cis-trans isomerase
Summary
Ara h 18 is a newly discovered peanut allergen component from the cyclophilin protein family with a molecular mass of 18.2 kDa (1). Biochemically, members of the cyclophilin protein family have the enzymatic function of peptidyl-prolyl cis-trans isomerases and are known for their high-affinity binding to the immunosuppressive agent cyclosporine A (2).  Cyclophilins are highly conserved and have been reported as IgE binding proteins in grass, tree, and weed pollen, several plant foods such as carrot, pumpkin, and tomato, as well as in several fungi and house dust mites (1), and are officially recognized allergens in several of those allergen sources (3).
Epidemiology
Worldwide distribution
Peanuts are consumed across the world and according to literature, peanut allergy affects approximately 2% of the population in Western nations, with data suggesting an increasing prevalence and incidence (4). Peanut allergy usually begins in childhood and persists throughout the affected individual’s lifetime (4); however, approximately 20% of young children develop tolerance (5).
In a study with 124 individuals, which were IgE positive to peanut but had a negative result (<0.35 kUA/l) to all peanut components available for specific IgE testing in clinical routine (Ara h 1-3, 6, 8-9), 35 samples (28%) were found to have IgE to Ara h 18, with a median level among positives of 10.6 kUA/l. While Ara h 18 was comparable to profilin regarding prevalence and magnitude of sensitization in this population, 13 of the samples were uniquely positive to Ara h 18 (1).
Environmental Characteristics
Source and tissue
Ara h 18 is a cyclophilin protein peanut allergen component.
The peanut (Arachis hypogaea) belongs to the legume family (Leguminosae) (6).
Clinical Relevance
The clinical patterns of peanut allergy, from sensitization to severe clinical symptoms, depend on the exposure routes and the physicochemical properties of the involved peanut proteins (7).
Primary sensitization to peanut is generally characterized by sensitization to the storage proteins Ara h 1, Ara h 2, Ara h 3 and/or Ara h 6 and sensitization to these components is often associated with severe allergic symptoms to peanut (8, 9).
Cyclophilins are highly cross-reactive. Cross-reactive pollen sensitization may cause a positive test result to peanut despite the absence of a primary peanut sensitization. Individuals with IgE antibodies only to pollen-associated targets are considered to have a low risk of a severe reaction to peanut (8).
Disease severity
Cyclophilins could carry a risk of severe symptoms and reactions, similar to Lipid Transfer Proteins (LTPs) and defensins, but lower than oleosins and storage proteins (Fig. 1) (7). However, their heat stability has not yet been elucidated. Inhibition results using grass pollen extract showed that Ara h 18 is probably not a primary sensitizer, but a cross-reactive determinant (1). 
Figure 1: Increased risk for severe symptoms and anaphylactic reactions from left to right (7).
Cross-reactive molecules
Cyclophilins are highly conserved proteins and extensive immunological cross-reactivity has been demonstrated between cyclophilins from different sources. Ara h 18 has a high sequence similarity to pollen cyclophilins from birch, olive and periwinkle (1). Sequence identity has been shown to correlate with immunological cross-reactivity and might have implications in the clinic (10).  
Molecular Aspects
Biochemistry
At the time of writing, 17 peanut allergens have been identified (7).
Ara h 18 has a molecular mass of 18.2 kDa and is a cyclophilin protein (1). Biochemically, members of the cyclophilin protein family have the enzymatic function of peptidyl-prolyl cis-trans isomerases and are known for their high-affinity binding to the immunosuppressive agent cyclosporine A (2). 
Cross-reactivity due to structural similarity
Ara h 18 is 88-91 % identical to cyclophilins in birch (Bet v 7), olive (Ole e 15) and periwinkle (Cat r 1) pollen (1).
Diagnostic Relevance
Ara h 18 is unlikely to be a primary sensitizer, but a cross-reactive determinant, binding IgE antibodies elicited by sensitization to another common allergen source, such as pollen. Ara h 18 may be helpful in explaining a positive peanut sensitization test result in the absence of IgE to currently available peanut components, which may elicit unjustified anxiety and unnecessary dietary avoidance/restriction (1).
Compiled By
Author: Dr. Merce Tena-Campos
Reviewer: Dr. Merima Mehic Chaveton & Ulrica G. Olsson
Last reviewed: 2023-07-17
References
  1. Mattsson L, Valcour A, Holmqvist M, Larsson H, Lidholm J. Cyclophilin - A novel cross-reactive determinant in peanut. Clin Exp Allergy. 2021;51(4):620-2.
  2. Stamnes MA, Rutherford SL, Zuker CS. Cyclophilins: a new family of proteins involved in intracellular folding. Trends Cell Biol. 1992;2(9):272-6.
  3. Sub-Committee WIAN. Allergen nomenclature  [cited 2023. Available from: www.allergen.org.
  4. Lieberman JA, Gupta RS, Knibb RC, Haselkorn T, Tilles S, Mack DP, et al. The global burden of illness of peanut allergy: A comprehensive literature review. Allergy. 2021;76(5):1367-84.
  5. Sicherer SH. Clinical update on peanut allergy. Ann Allergy Asthma Immunol. 2002;88(4):350-61; quiz 61-2, 94.
  6. Gepts P, Beavis WD, Brummer EC, Shoemaker RC, Stalker HT, Weeden NF, et al. Legumes as a model plant family. Genomics for food and feed report of the Cross-Legume Advances Through Genomics Conference. Plant Physiol. 2005;137(4):1228-35.
  7. Dramburg S, Hilger C, Santos AF, de Las Vecillas L, Aalberse RC, Acevedo N, et al. EAACI Molecular Allergology User's Guide 2.0. Pediatr Allergy Immunol. 2023;34 Suppl 28:e13854.
  8. Krogulska A, Wood RA. Peanut allergy diagnosis: Moving from basic to more elegant testing. Pediatr Allergy Immunol. 2020;31(4):346-57.
  9. Nicolaou N, Poorafshar M, Murray C, Simpson A, Winell H, Kerry G, et al. Allergy or tolerance in children sensitized to peanut: prevalence and differentiation using component-resolved diagnostics. J Allergy Clin Immunol. 2010;125(1):191-7 e1-13.
  10. Cadot P, Nelles L, Srahna M, Dilissen E, Ceuppens JL. Cloning and expression of the cyclophilin Bet v 7, and analysis of immunological cross-reactivity among the cyclophilin A family. Mol Immunol. 2006;43(3):226-35.