Contact Us
ImmunoCAP ISAC Test
Unparalleled Answers to Puzzling Allergies
These products may not be cleared for use in your country. Please contact your sales representative for information about specific product availability.
Determining the real causes of an allergic reaction may require several pieces of information. This is especially true when symptoms and case history are inconsistent, or the patient is multisensitized or shows unsatisfactory response to the treatment. In these puzzling cases, making the correct diagnosis may be both time-consuming and problematic.
The ImmunoCAP™ ISAC assay is a multiplex specific IgE test with allergen components to help healthcare providers resolve puzzling allergies.1,2
A test you can trust to provide unparalleled results
ImmunoCAP ISAC is the multiplex specific IgE test that combines clinically relevant composition of allergens, interpretation aids, broad documentation, and multiple years of experience—with only one drop of blood.*
Hidden Button for Modal Video
Clinically relevant selection of allergen components
112 allergen components from 48 different allergen sources have been thoroughly selected based on evidence in clinical literature for their potential to improve allergy diagnosis, assess risk for systemic reactions, and determine allergen(s) for successful allergen immunotherapy.1,2
Interpreted results provide the whole picture, at-a-glance
Results come in a structured report providing a comprehensive overview of a patient’s primary and cross-reactive sensitizations, as well as the clinical implication. This report helps simplify diagnosis of allergy and support clinical decision-making.
2009-03-13 Uppsala. Phadia. Isac. Foto: Markus Marcetic
Clinically validated and clinically proven
The ImmunoCAP ISAC assay has shown to provide robust performance, good reproducibility, and low variation demonstrated in a global validation study.3 More than 12 years of experience from research and clinical practice have resulted in more than 260 peer-reviewed publications.4
*30 µl of serum or plasma
ImmunoCAP ISAC tests
View Catalog
Article Number 81-1020-01
Product Type Test
Size 5 x 4 tests
Product Info
Explore the full ImmunoCAP portfolio.
View Portfolio
Find an instrument to run ImmunoCAP tests.
View Phadia Laboratory Systems
Get scientific information on whole allergens, allergen mixes, and allergen components.
Visit Allergen Encyclopedia
References
1.       Matricardi, P.M., et al., EAACI Molecular Allergology User's Guide. Pediatr Allergy Immunol, 2016. 27 Suppl 23: p. 1-250.
2.       Directions for use. ImmunoCAP ISAC E112i.
3.       van Hage, M., et al., Performance evaluation of ImmunoCAP(R) ISAC 112: a multi-site study. Clin Chem Lab Med, 2017. 55(4): p. 571-577.
4.       Thermo Fisher Scientific compiled list of publications. 20190520.