          • Proteins & Peptides ›
          • Eotaxin Proteins
          Gibco
          Mouse CCL11 (Eotaxin) Recombinant Protein, PeproTech®
          32 References
          View all (4) Eotaxin proteins
          Tech Support
          Cite Mouse CCL11 (Eotaxin) Recombinant Protein, PeproTech®
          {{$ctrl.translations['antibody.pdp.skuNotes.additionalInformation']}}:
          {{$ctrl.translations[$ctrl.isSkuNotesCollapsed ? 'antibody.pdp.skuNotes.viewMore' : 'antibody.pdp.skuNotes.viewLess']}}
          • Testing Data (1)
          Mouse CCL11 (Eotaxin) Protein in Bioactivity (Bioactivity)
          Group 53 Created with Sketch.
          Mouse CCL11 (Eotaxin) Protein in Bioactivity (Bioactivity)
          Group 53 Created with Sketch.
          FIGURE: 1 / 1
          Mouse CCL11 (Eotaxin) Protein (250-01-500UG) in Bioactivity
          Bioassay analysis of Mouse CCL11 (Eotaxin) Recombinant Protein, PeproTech® (Product # 250-01-1MG). {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product
          {{$ctrl.videoDesc}}
          {{$ctrl.videoLongDesc}}
          Mouse CCL11 (Eotaxin) Protein in Bioactivity (Bioactivity)
          Product Details
          250-01-500UG
          Applications
          Tested Dilution
          Publications
          Control (Ctrl)
          Assay-dependent
          -
          In vitro Assay (In vitro)
          -
          View 21 publications 21 publications
          Miscellaneous PubMed (Misc)
          -
          View 9 publications 9 publications
          Western blot control (WB Ctrl)
          Assay-dependent
          -
          ELISA standard (ELISA Std)
          Assay-dependent
          View 2 publications 2 publications
          Bioactivity (Bioactivity)
          Assay-dependent
          -
          Product Specifications
          Species
          Mouse
          Published species
          Human, Mouse
          Expression System
          E. coli
          Amino acid sequence
          HPGSIPTSCC FIMTSKKIPN TLLKSYKRIT NNRCTLKAIV FKTRLGKEIC ADPKKKWVQD ATKHLDQKLQ TPKP
          Molecular weight
          8.4 kDa
          Class
          Recombinant
          Type
          Protein
          Purity
          ≥ 98% by SDS-PAGE gel and HPLC analyses.
          Endotoxin concentration
          <1 EU/µg
          Activity
          Murine Eotaxin was found to induce chemotaxis of purified eosinophils at concentrations ranging between 10-100 ng/ml.
          Conjugate
          Unconjugated Unconjugated Unconjugated
          Form
          Lyophilized
          Amount
          500 µg
          Purification
          purified
          Contains
          no preservative
          Storage conditions
          -20°C
          Shipping conditions
          Ambient
          Product Specific Information
          Recombinant Murine Eotaxin is an 8.4 kDa protein containing 74 amino acid residues, including the four highly conserved cysteine residues present in CC chemokines.
          This product is shipped at ambient temperature. For storage, handling and reconstitution information, please see the lot-specific Certificate of Analysis
          Target Information
          This gene is one of several Cys-Cys (CC) cytokine genes clustered on the q-arm of chromosome 17. Cytokines are a family of secreted proteins involved in immunoregulatory and inflammatory processes. The CC cytokines are proteins characterized by two adjacent cysteines. The cytokine encoded by this gene displays chemotactic activity for eosinophils, but not mononuclear cells or neutrophils. This eosinophil specific chemokine assumed to be involved in eosinophilic inflammatory diseases such as atopic dermatitis, allergic rhinitis, asthma and parasitic infections.
          Bioinformatics
          Protein Aliases: C-C motif chemokine 11; chemokine (C-C motif) ligand 11; Eosinophil Chemotactic Factor; eosinophil chemotactic protein; Eotaxin 1; Eotaxin1; MGC22554; small chemokine (C-C motif) ligand 11; small-inducible cytokine A11; unnamed protein product
          View more View less
          Gene Aliases: eotaxin; Scya11
          View more View less
          UniProt ID: (Mouse) P48298
          View more View less
          Entrez Gene ID: (Mouse) 20292
          Contact tech support
          オンラインオーダー Plus Icon Minus Icon
          • オンラインオーダー可能な製品
          • オンラインオーダーに関する個人情報の取扱いについて
          • 特定商取引法に基づく表記
          • 返品について
          • 表示価格について
          • 配送料について
          サポート Plus Icon Minus Icon
          • FAQ、サポート情報
          • お問い合わせ一覧
          • サポートセンター
          • ドキュメント、証明書
          • 研究情報公開
          • Webサイトの問題を報告​
          リソース Plus Icon Minus Icon
          • ラーニングセンター
          • キャンペーン
          • イベント、セミナー
          • Blog
          • 企業情報
          • Social Responsibility (CSR)
          • お客さま体験の向上
          • 採用情報 採用情報
          • ニュース
          • 商標
          • ポリシーと通知
          • PPD
          • 標準販売条件
          • プライバシー通知
          Japan flag icon
          Japan
          Your items have has been added!
          Host server : magellan-search-blue-7fb9f846c9-xq5gc:80/100.66.76.64:80.
          git-commit: 8207b5afa5c305bd40e0a4f0b9b53fd46f2a8829
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.59.1-2026.09.91-1.0