Search
Search

AT1G16570| Gene Symbol: AT1G16570 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001123825.1 | NP_001117297.1 | 3 - 4 | 178 | 85 | |
| NM_101521.3 | NP_173105.1 | 3 - 4 | 178 | 85 | ||
| GenBank mRNA | AY128379.1 | - | 3 - 4 | 171 | 85 | |
| BT008762.1 | - | 2 - 3 | 126 | 85 | ||
| KJ139004.1 | - | 2 - 3 | 126 | 85 | ||
Set Membership: |
|