Search
Search

VPS46.1| Gene Symbol: VPS46.1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_101635.5 | NP_173215.1 | 1 - 2 | 497 | 65 | |
| GenBank mRNA | AK118350.1 | - | 1 - 2 | 425 | 65 | |
| AY085606.1 | - | 1 - 2 | 421 | 65 | ||
Set Membership: |
|