Search
Search

GASA5| Gene Symbol: GASA5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_111158.4 | NP_566186.1 | 2 - 3 | 281 | 59 | |
| GenBank mRNA | BT002786.1 | - | 2 - 3 | 155 | 59 | |
| BT004355.1 | - | 2 - 3 | 114 | 59 | ||
Set Membership: |
|