Search
Search

LOL2| Gene Symbol: LOL2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001341506.1 | NP_001329633.1 | 2 - 3 | 274 | 104 | |
| NM_118281.4 | NP_193892.1 | 2 - 3 | 367 | 104 | ||
| GenBank mRNA | BT011760.1 | - | 2 - 3 | 367 | 104 | |
| BT012384.1 | - | 2 - 3 | 274 | 104 | ||
Set Membership: |
|