Search
Search

AT4G21660| Gene Symbol: AT4G21660 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001203860.1 | NP_001190789.1 | 11 - 12 | 1298 | 66 | |
| NM_001341508.1 | NP_001328547.1 | 10 - 11 | 1603 | 66 | ||
| NM_118286.4 | NP_193897.2 | 10 - 11 | 1321 | 66 | ||
| GenBank mRNA | BT002013.1 | - | 10 - 11 | 1228 | 66 | |
| BT010557.1 | - | 10 - 11 | 1186 | 66 | ||
Set Membership: |
|