Search
Search

GA2OX2| Gene Symbol: GA2OX2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001036035.2 | NP_001031112.1 | 1 - 2 | 532 | 89 | |
| NM_102743.3 | NP_174296.1 | 1 - 2 | 723 | 89 | ||
| GenBank mRNA | AJ132436.1 | - | 1 - 2 | 505 | 89 | |
| BT002987.1 | - | 1 - 2 | 532 | 89 | ||
| BT004464.1 | - | 1 - 2 | 411 | 89 | ||
Set Membership: |
|