Search
Search

AT1G30280| Gene Symbol: AT1G30280 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001332885.1 | NP_001322470.1 | 1 - 2 | 481 | 97 | |
| NM_001332886.1 | NP_001322469.1 | 1 - 2 | 481 | 97 | ||
| NM_102767.4 | NP_174319.1 | 1 - 2 | 579 | 97 | ||
| GenBank mRNA | AY070092.1 | - | 1 - 2 | 579 | 97 | |
| AY117364.1 | - | 1 - 2 | 481 | 97 | ||
Set Membership: |
|