Search
Search

CCC1| Gene Symbol: CCC1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001332900.1 | NP_001323337.1 | 10 - 11 | 1751 | 92 | |
| NM_102781.3 | NP_174333.2 | 12 - 13 | 2051 | 92 | ||
| NM_179400.1 | NP_849731.1 | 12 - 13 | 2020 | 92 | ||
| NM_179401.2 | NP_849732.1 | 12 - 13 | 2051 | 92 | ||
| GenBank mRNA | AK226602.1 | - | 12 - 13 | 2006 | 92 | |
| AM113986.1 | - | 12 - 13 | 2021 | 92 | ||
Set Membership: |
|