Search
Search

CKB4| Gene Symbol: CKB4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001337093.1 | NP_001323623.1 | 1 - 2 | 453 | 96 | |
| NM_130032.3 | NP_181996.1 | 1 - 2 | 453 | 96 | ||
| NM_180090.2 | NP_850421.1 | 1 - 2 | 453 | 96 | ||
| GenBank mRNA | AY052265.1 | - | 1 - 2 | 410 | 96 | |
| AY060523.1 | - | 1 - 2 | 222 | 96 | ||
| AY062585.1 | - | 1 - 2 | 413 | 96 | ||
| AY088075.1 | - | 1 - 2 | 413 | 96 | ||
| AY093344.1 | - | 1 - 2 | 222 | 96 | ||
Set Membership: |
|