Search
Search

AT2G45460| Gene Symbol: AT2G45460 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001202821.1 | NP_001189750.1 | 9 - 10 | 1454 | 146 | |
| NM_001202822.2 | NP_001189751.1 | 9 - 10 | 1487 | 146 | ||
| NM_001337132.1 | NP_001325090.1 | 9 - 10 | 1452 | 146 | ||
| NM_180106.1 | NP_850437.1 | 9 - 10 | 1303 | 146 | ||
Set Membership: |
|