Search
Search

AT3G07140| Gene Symbol: AT3G07140 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_111594.4 | NP_566299.1 | 2 - 3 | 1589 | 128 | |
| NM_180199.2 | NP_850530.1 | 2 - 3 | 1496 | 128 | ||
| GenBank mRNA | AX765858.1 | - | 2 - 3 | 1415 | 128 | |
| AY050884.1 | - | 2 - 3 | 1456 | 128 | ||
| AY133829.1 | - | 2 - 3 | 1415 | 128 | ||
Set Membership: |
|