Search
Search

KINESIN-13A| Gene Symbol: KINESIN-13A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_112536.3 | NP_188285.1 | 10 - 11 | 1866 | 97 | |
| NM_180267.2 | NP_850598.1 | 10 - 11 | 1844 | 97 | ||
| GenBank mRNA | AK226569.1 | - | 10 - 11 | 1721 | 97 | |
| AY056129.1 | - | 10 - 11 | 1702 | 97 | ||
Set Membership: |
|