Search
Search

FRK1| Gene Symbol: FRK1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_127476.2 | NP_179509.1 | 10 - 11 | 2025 | 109 | |
| GenBank mRNA | AF486619.1 | - | 10 - 11 | 2025 | 109 | |
| FJ708695.1 | - | 10 - 11 | 1974 | 109 | ||
Set Membership: |
|