Search
Search

SYN1| Gene Symbol: SYN1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_120631.2 | NP_196168.1 | 19 - 20 | 1921 | 74 | |
| NM_180442.2 | NP_850773.1 | 20 - 21 | 1921 | 74 | ||
| GenBank mRNA | AF080619.1 | - | 19 - 20 | 1924 | 74 | |
| AF080620.1 | - | 20 - 21 | 1921 | 74 | ||
| AJ238306.1 | - | 19 - 20 | 1828 | 74 | ||
Set Membership: |
|