Search
Search

ACR12| Gene Symbol: ACR12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001203298.1 | NP_001190227.1 | 1 - 2 | 276 | 76 | |
| NM_120556.3 | NP_196094.1 | 1 - 2 | 346 | 76 | ||
| GenBank mRNA | AY072401.1 | - | 1 - 2 | 276 | 76 | |
| AY114702.1 | - | 1 - 2 | 187 | 76 | ||
| JF797177.1 | - | 1 - 2 | 187 | 76 | ||
Set Membership: |
|