Search
Search

NAPRT2| Gene Symbol: NAPRT2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001335851.1 | NP_001323701.1 | 1 - 2 | 359 | 73 | |
| NM_127906.6 | NP_179923.2 | 1 - 2 | 360 | 73 | ||
| GenBank mRNA | BT001932.1 | - | 1 - 2 | 144 | 73 | |
Set Membership: |
|