Search
Search

NPH4| Gene Symbol: NPH4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_122080.3 | NP_568400.2 | 10 - 11 | 1758 | 60 | |
| NM_180715.1 | NP_851046.1 | 10 - 11 | 1758 | 60 | ||
| NM_180716.3 | NP_851047.1 | 10 - 11 | 1821 | 60 | ||
| GenBank mRNA | AY669789.1 | - | 10 - 11 | 1146 | 60 | |
Set Membership: |
|