Search
Search

P5CS2| Gene Symbol: P5CS2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001125374.1 | NP_001118846.1 | 4 - 5 | 437 | 76 | |
| NM_115419.5 | NP_191120.2 | 4 - 5 | 639 | 76 | ||
| GenBank mRNA | AY091766.1 | - | 4 - 5 | 437 | 76 | |
| Y09355.1 | - | 407 | 76 | |||
Set Membership: |
|