Search
Search

PAO3| Gene Symbol: PAO3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_115767.3 | NP_191464.1 | 3 - 4 | 690 | 71 | |
| GenBank mRNA | AY065025.1 | - | 3 - 4 | 672 | 71 | |
| AY143905.1 | - | 3 - 4 | 354 | 71 | ||
Set Membership: |
|