Search
Search

VSR2| Gene Symbol: VSR2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001202708.1 | NP_001189637.1 | 9 - 10 | 1612 | 98 | |
| NM_128582.2 | NP_180588.1 | 9 - 10 | 1615 | 98 | ||
| GenBank mRNA | DQ446580.1 | - | 9 - 10 | 1481 | 98 | |
Set Membership: |
|