Search
Search

EML3| Gene Symbol: EML3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001343254.1 | NP_001332263.1 | 5 - 6 | 1011 | 72 | |
| NM_001343255.1 | NP_001332264.1 | 5 - 6 | 1146 | 72 | ||
| NM_121305.4 | NP_196806.2 | 5 - 6 | 1061 | 72 | ||
Set Membership: |
|