Search
Search

GASA4| Gene Symbol: GASA4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001085118.2 | NP_001078587.1 | 2 - 3 | 298 | 92 | |
| NM_121527.4 | NP_197027.1 | 3 - 4 | 344 | 92 | ||
| GenBank mRNA | AF360199.1 | - | 3 - 4 | 257 | 92 | |
| AY040048.1 | - | 3 - 4 | 143 | 92 | ||
| AY057601.1 | - | 3 - 4 | 257 | 92 | ||
Set Membership: |
|