Search
Search

AT5G19070| Gene Symbol: AT5G19070 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001343586.1 | NP_001331805.1 | 5 - 6 | 724 | 56 | |
| NM_121912.3 | NP_197408.2 | 5 - 6 | 805 | 56 | ||
| GenBank mRNA | AK228918.1 | - | 5 - 6 | 626 | 56 | |
| BT014970.1 | - | 5 - 6 | 642 | 56 | ||
| BT020544.1 | - | 5 - 6 | 395 | 56 | ||
Set Membership: |
|