Search
Search

SYCO ARATH| Gene Symbol: SYCO ARATH | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001084517.1 | NP_001077986.1 | 2 - 3 | 457 | 149 | |
| NM_128673.5 | NP_565717.2 | 2 - 3 | 465 | 145 | ||
Set Membership: |
|